MicroSEQ™ ID 16S rDNA Full Gene Library v2.1 - FAQs
MicroSEQ™ ID 16S rDNA Full Gene Library v2.1 - FAQs
View additional product information for MicroSEQ™ ID 16S rDNA Full Gene Library v2.1 - FAQs (A57315)
7 product FAQs found
What are the operating system requirements for MicroSEQ ID 16S rDNA Full Gene Library v2.1?
The operating system requirements for MicroSEQ ID 16S rDNA Full Gene Library v2.1 are Windows 10, Enterprise LTSC 2018 Microsoft Corporation, 64 bit OS (Operating System), x64 Based Processor, Version-1809.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the difference between MicroSEQ ID 16S rDNA Full Gene Library v2.0 and v2.1?
MicroSEQ ID 16S rDNA Full Gene Library v2.1 contains identical content to the previous generation library, MicroSEQ ID 16S rDNA Full Gene Library v2.0. MicroSEQ ID 16s rDNA Full Gene Library v2.1 is compatible with MicroSEQ ID Microbial Identification Software v4.0 whereas MicroSEQ ID 16s rDNA Full Gene Library v2.0 is compatible with MicroSEQ ID Microbial Identification Software v3.1.3.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the browser compatibility requirement for MicroSEQ ID 16S rDNA Full Gene Library v2.1?
MicroSEQ ID 16S rDNA Full Gene Library v2.1 is compatible with Google Chrome v108.0.5359.73.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which Windows OS (Operating System) is MicroSEQ ID Microbial Identification Software v4.0 compatible with?
MicroSEQ ID Microbial Identification Software v4.0 is compatible with 64-bit Windows 10 OS.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What are the computer requirements for MicroSEQ ID Microbial Identification Software v4.0?
The computer requirements for MicroSEQ ID Microbial Identification Software v4.0 are:
- Intel Core i7-8700 CPU at 3.20 Ghz, with 16 GB of RAM and a minimum of 465 GB hard disk space
- WIN 10 IoT 64-bit
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
Which MicroSeq ID (MSID) library versions is MicroSEQ ID Microbial Identification Software v4.0 compatible with?
MicroSEQ ID Microbial Identification Software v4.0 is compatible with MicroSEQ ID 16S rDNA Full Gene Library v2.1, MicroSEQ ID Fungal Gene Library v2022, MicroSEQ ID 16S rDNA 500 Library v2022, and MicroSEQ ID 16s rDNA 500 Supplemental Library v2022.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.
What is the product SKU for MicroSEQ ID 16S rDNA Full Gene Library v2.1?
The product SKU for MicroSEQ ID 16S rDNA Full Gene Library v2.1 is Cat. No. A57315.
Find additional tips, troubleshooting help, and resources within our Capillary Electrophoresis Software Support Center.