E-Gel™ EX Mix Pack
Invitrogen™

E-Gel™ EX Mix Pack

The E-Gel EX Mix Pack allows researchers to try all of our standard format E-Gels EX agarose gels. These pre-cast자세히 알아보기
Have Questions?
카탈로그 번호수량
A5734310 Gels
카탈로그 번호 A57343
제품 가격(KRW)
305,000
온라인 행사
Ends: 31-Mar-2026
358,000
할인액 53,000 (15%)
Each
수량:
10 Gels
제품 가격(KRW)
305,000
온라인 행사
Ends: 31-Mar-2026
358,000
할인액 53,000 (15%)
Each
The E-Gel EX Mix Pack allows researchers to try all of our standard format E-Gels EX agarose gels. These pre-cast agarose gels are designed for ultrasensitive and convenient DNA sample electrophoresis in as little as eight minutes.

Benefits of using E-Gels EX gels include:
High sensitivity—detect 1 ng of sample DNA
Rapid analysis—complete DNA sample separation in eight minutes
Easy sample access—openable cassette design allows easy in-gel access for convenient DNA sample excision or transfer to membranes
Instantaneous setup—precast E-Gel agarose gels are ready for use and do not require gel preparation or additional buffers
No buffers—E-Gel agarose gels do not require buffers to run

The box contains:
• E-Gel EX Agarose Gels, 1%
• E-Gel EX Agarose Gels, 2%
• E-Gel EX Double Comb Agarose Gels, 1%
• E-Gel EX Double Comb Agarose Gels, 2%
• E-Gel SizeSelect II Agarose Gels, 2%

Running and imaging devices
E-Gels EX agarose gels require the E-Gel Power Snap or E-Gel Power Snap Plus electrophoresis systems for gel running and viewing. The systems consist of an electrophoresis device and a camera for fast and convenient E-Gel agarose gel separation and analysis.

For Research Use Only.
사양
수량10 Gels
멸균Non-sterile
형식Precast Gel
젤 유형E-Gel
분리 유형Electrophoresis
다음의 분리DNA
11-well
Unit SizeEach