CTS™ OpTmizer™ One SFM, BPC format, 1 L - FAQs
CTS™ OpTmizer™ One SFM, BPC format, 1 L - FAQs
View additional product information for CTS™ OpTmizer™ One SFM - FAQs (A5757204, A5757203, A5757202, A5757201)
6 product FAQs found
What is CTS?
The Gibco Cell Therapy Systems (CTS) portfolio of cell and gene therapy products are GMP manufactured, safety tested, and backed by regulatory documentation to support your transition from discovery through clinical and commercial manufacturing. Through our CTS solutions, we are committed to helping customers streamline therapeutic development, minimize risk, and ease the burden on their quality systems. Learn more here.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does CTS OpTmizer One SFM work in bioreactors?
Yes. CTS OpTmizer One SFM offers excellent performance in a range of culture vessels, from static to dynamic bioreactors.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Do I need to add serum to CTS OpTmizer One SFM?
No. CTS OpTmizer One SFM was designed to be used without serum or serum replacement supplementation.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does CTS OpTmizer One SFM contain protein?
Yes, all protein within CTS OpTmizer One SFM is recombinant.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Does CTS OpTmizer One SFM contain L-glutamine?
No. The most consistent and reproducible results are seen when L-glutamine is added to CTS OpTmizer One SFM at the point of use.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Is CTS OpTmizer One SFM an animal-origin-free (AOF) version of legacy CTS OpTmizer SFM?
No. CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium. It is not a kit like the legacy CTS OpTmizer SFM, and does not require mixing of separate basal medium and supplement components to create complete medium.
CTS OpTmizer One SFM does not contain or use in the manufacturing processes any primary or secondary raw materials that are derived directly from animal (including human) tissue or body fluid.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.