Zeba™ Spin Desalting Columns and Plates, 40K MWCO, 75 μL–10 mL
Thermo Scientific™

Zeba™ Spin Desalting Columns and Plates, 40K MWCO, 75 μL–10 mL

The new and enhanced Zeba Spin and Micro Spin Desalting columns and plates are designed for efficient desalting and buffer exchanging proteins with a molecular weight >40 kDa and removing small molecules less than 2 kDa. They provide exceptional desalting and protein recovery performance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySample Volume (Metric)
A577672 plates20 to 100 μL
A400024715 Columns4 to 10 mL
A4000247610 Columns4 to 10 mL
A5775625 columns5 to 14 μL
A5775850 columns5 to 14 μL
A5775925 columns50 to 150 μL
A5776050 columns50 to 150 μL
A577615 columns300 to 800 μL
A5776225 columns300 to 800 μL
A577635 columns500 to 2000 μL
A5776425 columns500 to 2000 μL
A577655 columns1000 to 4000 μL
A5776625 columns1000 to 4000 μL
A577684 plates20 to 100 μL
Catalog number A57767
Price (USD)
354.65
Online exclusive
377.00
Save 22.35
Each
Quantity:
2 plates
Sample Volume (Metric):
20 to 100 μL
Price (USD)
354.65
Online exclusive
377.00
Save 22.35
Each
Thermo Scientific Zeba spin and micro spin desalting columns and plates are designed for efficient desalting and buffer exchange of protein samples. They contain a proprietary size-exclusion chromatography resin with a robust bead structure that withstands centrifugal force and helps produce better flow characteristics and increased efficiency in removing low molecular weight protein contaminants from samples. The Zeba resin performs well even with significantly diluted protein samples and eliminates over 95% of contaminants without causing protein loss.

These Zeba spin and micro spin desalting columns and plates are designed for desalting and buffer exchange of proteins with a molecular weight >40 kDa and removal of small molecules less than 2 kDa. They provide exceptional desalting and protein recovery performance, with expanded capacity to accommodate up to 10 mL of sample volume.

Features of Zeba spin and micro spin desalting columns include:

  • High-performing resin—exceptional protein recovery at lower protein concentrations
  • Fast and efficient—eliminate sample dilution, fraction collection, and the need for expensive chromatography systems that take time to set up and can only process one sample at a time
  • Easy to use—no cumbersome column preparation or equilibration
  • Flexible formats—available in spin columns and 96-well spin plates for a range of sample volumes (2 μL to 10 mL)

Zeba Desalting Columns

Convenient, ready-to-use polypropylene columns are the ideal solution for quick and straightforward desalting and buffer exchange of protein samples in a centrifuge format. The pre-dispensed spin columns are offered in various resin fills (75 μL, 0.5 mL, 2 mL, 5 mL, 10 mL, 20mL). These options make it simple to handle sample volumes between 2 μL and 10 mL.

Zeba Spin Desalting Plates

These plates provide trouble-free high-throughput desalting and buffer exchange for sample volumes ranging from 20 to 100 μL. They require no resin dispensing or hydration and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 μL) in five minutes. Separating macromolecules, such as proteins, from small contaminants is often done through routine methods, including gel filtration chromatography and buffer exchange. These methods are commonly used during protein purification workflows to remove small contaminants like salts, excess labels, and cross-linking reagents from samples before analysis.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
DisposableSingle-use
Product TypeSpin Desalting Plate
Purification TargetBuffer Exchange, Protein
Quantity2 plates
Column TypeSize-exclusion, Proprietary Resin
Format96-well Filter Plate
MWCO40 kDa
Product LineZeba
Sample Volume (Metric)20 to 100 μL
Unit SizeEach

Frequently asked questions (FAQs)

Can Zeba Desalting columns, plates, and cartridges be autoclaved?

Yes, Zeba Desalting columns, plates, and cartridges can be autoclaved at 115 degrees C for 30 min.

Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.

What reagents and conditions are Zeba Desalting columns, plates, and cartridges compatible with?

Zeba Desalting columns, plates, and cartridges are compatible with a range of reagents and conditions. They have been confirmed to be compatible with the following:
- pH range of 3-10
- Mild oxidants/reductants
- Chaotropes (stable in 8 M urea and 6 M guanidine HCl)
- Salts
- Alcohols up to 20%
- Organic solvents such as dimethyl sulfoxide (DMSO) and dimethylformamide (DMF) (we recommend using a step-gradient of increasing concentration of the organic solvent during column equilibration)

Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.

Can I use Zeba Desalting plates with a vacuum manifold, and if so, are there any specific considerations or limitations to keep in mind?

Yes, Zeba Desalting plates can be used with a vacuum manifold. Unfortunately, we do not offer a protocol for the same. It is important to note that the pressure settings of the vacuum manifold will need to be optimized for each instrument as the pressure apparatus may differ. Additionally, it has been reported that protein recovery with Zeba Desalting plates using a vacuum manifold may be lower compared to the traditional centrifugation workflow.

Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.