EasyPep™ 96 Micro MS Sample Prep Kit, 1–10 μg, 96 reactions - FAQs
EasyPep™ 96 Micro MS Sample Prep Kit, 1–10 μg, 96 reactions - FAQs
View additional product information for EasyPep™ MS Sample Prep Kits - FAQs (A45733, A45734, A40006, A45735, A57865, A57864)
1 product FAQs found
Are the EasyPep MS Sample Prep Kits compatible with vacuum pumps/dry-down devices?
As per protocol, peptide samples need to be dried down prior to LC-MS analysis. This step can be performed using vacuum pumps/dry-down devices without any harm caused to the device by the components present in the elution buffer within the EasyPep MS Sample Prep Kits.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.