Sodium Chloride (NaCl) Solutions, 0.1–2.0 M
Gibco™

Sodium Chloride (NaCl) Solutions, 0.1–2.0 M

Gibco Sodium Chloride (NaCl) solutions are sterile-filtered, packaged in bioprocess containers, and ready-to-use. Sourcing NaCl solutions can help streamline your buffer preparation allowing you to focus on additional critical steps in your biopharmaceutical processes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackagingConcentration
A58029015 LSample0.5 M
A57922015 LSample0.1 M
A579220220 LTote0.1 M
A5792203200 LDrum0.1 M
A58028015 LSample0.2 M
A580280220 LTote0.2 M
A5802803200 LDrum0.2 M
A580290220 LTote0.5 M
A5802903200 LDrum0.5 M
A58030015 LSample1.0 M
A580300220 LTote1.0 M
A5803003200 LDrum1.0 M
A58031015 LSample2.0 M
A580310220 LTote2.0 M
A5803103200 LDrum2.0 M
Catalog number A5802901
Price (USD)
-
Quantity:
5 L
Packaging:
Sample
Concentration:
0.5 M
We mix compendial-grade sodium chloride (NaCl) with the same bulk water used in our Gibco Water for Injection (WFI) for Cell Culture to produce solutions that meet the most stringent expectations of the pharmaceutical, biopharma, and diagnostic industries. NaCl solutions are manufactured to cGMP (21CFR820) and ISO 13485 standards, promoting consistency and convenience for every stage of the bioprocessing pipeline.

Product features:
• Multiple concentrations available (0.1 M, 0.2 M, 0.5 M, 1.0 M, 2.0 M)
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Established products with set specifications are ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification

User benefits:
• Eases the time requirements dedicated for in-house buffer preparation
• Mitigates space required for equipment storage
• Streamlines in-house chromatography processes related to formulation, hydration, testing, and validation
• Meets workflow needs for all stages of biologic development and manufacturing

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
CAS7647-14-5
Certifications/ComplianceGMP - 21 CFR 820, ISO 13485
Chemical Name or MaterialSodium Chloride
ColorClear
Endotoxin Level≤ 1.0 EU/mL
Formulation0.5 M NaCl
Osmolality880–980 mOsm/kg
PackagingSample
Shelf Life12 Months
SterilitySterile-filtered
Sterilization Method0.2 μm Sterile-filtered
Concentration0.5 M
For Use With (Application)Bioproduction
GradeCompendial
Molecular Weight (g/mol)58.44
Physical FormLiquid
Quantity5 L
Solution TypeBuffer
pH4.5–8.0
Unit SizeEach