TrueMark™ Enteric Parasitic Select Panel Combo Kit
TrueMark™ Enteric Parasitic Select Panel Combo Kit
Applied Biosystems™

TrueMark™ Enteric Parasitic Select Panel Combo Kit

The TrueMark Enteric Parasitic Select Panel Combo Kit includes the TrueMark Enteric Parasitic Select Panel and TrueMark Infectious Disease 1‐StepRead more
Have Questions?
Catalog number A58077
Price (USD)/ Each
3,600.00
Add to cart
Price (USD)/ Each
3,600.00
Add to cart
TrueMark™ Enteric Parasitic Select Panel Combo Kit
Catalog numberA58077
Price (USD)/ Each
3,600.00
-
Add to cart

The TrueMark Enteric Parasitic Select Panel Combo Kit includes the TrueMark Enteric Parasitic Select Panel and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark Enteric Parasitic Select Amplification Control (offered separately), the kit can be used to quickly evaluate and research up to 200 samples consisting of extracted nucleic acid from stool specimens.

The kit is recommended for use with the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block). It includes primers and probe sets specific to the following targets:
• Cryptosporidium spp.
Entamoeba histolytica
Giardia lamblia
Cyclospora cayetanensis
Bacillus atrophaeus (used as a process control)

Features of the TrueMark Enteric Parasitic Select Panel Combo Kit include:
• Detects and differentiates targets found in enteric parasitic infections
• Affordable and scalable—add to existing infrastructure to expand research menu while maintaining low operational costs
• Ready-to-use off-the-shelf solution—seamlessly integrate test into existing workflow

The TrueMark Enteric Parasitic Select Panel Combo Kit includes the following components:
TrueMark Enteric Parasitic Select Panel—multiplexed assay that contains primer and probe sets
TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)—ready-to-use PCR mix, including polymerase, dNTPs, salt, and buffer

Use this kit with:
TrueMark Enteric Parasitic Select Amplification Control—plasmid control containing target-specific templates

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Research Use Only.
Specifications
Detection MethodqPCR
FormLiquid
PCR MethodMultiplex qPCR, qPCR, RT-PCR
Reaction Speed120 min.
Quantity200 reactions
For Use With (Application)Pathogen Detection, Real Time PCR (qPCR), RT-PCR
No. of Reactions200 reactions
Unit SizeEach