TrueMark™ Enteric Norovirus Select Panel Combo Kit
Applied Biosystems™

TrueMark™ Enteric Norovirus Select Panel Combo Kit

The TrueMark Enteric Norovirus Select Panel Combo Kit includes the TrueMark Enteric Norovirus Select Panel and TrueMark Infectious Disease 1‐StepRead more
Have Questions?
Catalog NumberQuantity
A58078200 reactions
Catalog number A58078
Price (MXN)
-
Quantity:
200 reactions

The TrueMark Enteric Norovirus Select Panel Combo Kit includes the TrueMark Enteric Norovirus Select Panel and TrueMark Infectious Disease 1‐Step Multiplex Master Mix for simplified ordering. Together with the TrueMark Enteric Norovirus Select Amplification Control (offered separately), the kit can be used to quickly evaluate and research up to 200 samples consisting of extracted nucleic acid from stool specimens.

The kit is recommended for use with the QuantStudio 5 Real-Time PCR System (with 96-well, 0.2-mL block). It includes primers and probe sets specific to the following targets:
• Norovirus GI
• Norovirus GII
Bacillus atrophaeus (used as a process control)

Features of the TrueMark Enteric Norovirus Select Panel Combo Kit include:
• Detects and differentiates targets found in enteric norovirus infections
• Affordable and scalable—add to existing infrastructure to expand research menu while maintaining low operational costs
• Ready-to-use off-the-shelf solution—seamlessly integrate test into existing workflow

The TrueMark Enteric Norovirus Select Panel Combo Kit includes the following components:
TrueMark Enteric Norovirus Select Panel—multiplexed assay that contains primer and probe sets
TrueMark Infectious Disease 1‐Step Multiplex Master Mix (No ROX)—ready-to-use PCR mix, including polymerase, dNTPs, salt, and buffer

Use this kit with:
TrueMark Enteric Norovirus Select Amplification Control—plasmid control containing target-specific templates

For Research Use Only.
Specifications
No. of Reactions200 reactions
Quantity200 reactions
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Real Time PCR (qPCR), RT-PCR
FormLiquid
PCR MethodMultiplex qPCR, qPCR, RT-PCR
Reaction Speed120 min.
Unit SizeEach