TaqPath™ Menu ∣ GeneProof™ Neisseria gonorrhoeae PCR Kit
TaqPath™ Menu ∣ GeneProof™ Neisseria gonorrhoeae PCR Kit
Applied Biosystems™

TaqPath™ Menu ∣ GeneProof™ Neisseria gonorrhoeae PCR Kit

The TaqPath Menu | GeneProof Neisseria gonorrhoeae PCR Kit is a real-time PCR testing solution that enables dual target detectionRead more
Have Questions?
Catalog NumberQuantity
A58124100 reactions kit
Catalog number A58124
Price (CLP)
1.249.024
Each
Add to cart
Quantity:
100 reactions kit
Price (CLP)
1.249.024
Each
Add to cart

The TaqPath Menu | GeneProof Neisseria gonorrhoeae PCR Kit is a real-time PCR testing solution that enables dual target detection of the 16S rRNA gene and the porA pseudogene. The kit is approved for use on the QuantStudio 3/5 real-time PCR systems (96-well, 0.2 mL block) and the Applied Biosystems 7300/7500 real-time PCR systems (96-well).

Features of the TaqPath Menu | GeneProof Neisseria gonorrhoeae PCR Kit include:
• High Sensitvity secured by targetingmulticopy sequence of 16S rRNA gene
• Reliable detection of porA mutant
• Minimizes contamination—the master mix contains uracil-DNA glycosylase (UNG) and dUTPs, reducing carryover contamination
• Compatible with a wide range of real-time PCR devices

The TaqPath Menu | GeneProof Neisseria gonorrhoeae PCR Kit enables the qualitative diagnostics and aid to diagnosis of Neisseria gonorrhoeae from sperm, swab, and urine by polymerase chain reaction (PCR) method. It is designed for professional use in laboratories with trained staff.


Access the Instructions for Use (IFU) approved for your country

Explore more sexually transmitted infections solutions.
Available in select countries only.
Specifications
Certification ComplianceCE2797 IVD / Regulation (EU) 2017/746
CompatibilityRegularly tested by QCMD and Instand e.V. External Quality Assessment Panels
Control TemplatePCR Inhibition and DNA extraction efficiency control
For Use With (Equipment)QuantStudio™ 3/5 Real-Time PCR System, Applied Biosystems 7300/7500 Real-Time PCR System, StepOne™/StepOne Plus™ Real-Time PCR System, AMPLilab Real-Time PCR System, AriaMx Real-Time PCR System, BioQuant-96 Real-Time PCR System, CFX Connect™/CFX96™ Dx Real-Time PCR Detection System. CFX Opus 96 Real-Time PCR System, croBEE Real-Time PCR System, Gentier 96E/96R Real-Time PCR System, LightCycler™ 480, LineGene 9600/9600 Plus, Mic qPCR Cycler, Rotor-Gene 3000/6000/Q, SLAN™ Real-Time PCR System
No. of Reactions100 Reactions
Product LineTaqPath
Quantity100 reactions kit
Sample TypeDNA extracted from swab (urethral, cervical, vaginal), urine, sperm
SensitivityAnalytical: 0.109 cp/μL (Neisseria gonorrhoeae)
Target16S rRNA gene and porA pseudogene
Target SpecificityAnalytical: 100% (N. gonorhoeae, including mutants in porA pseudogene)
TechniqueQualitative Analysis
Label or DyeFAM
PCR MethodqPCR
Unit SizeEach
Contents & Storage
• Master mix
• Positive Control

Store at –20 ± 5°C.