Search
| Número de catálogo | Cantidad |
|---|---|
| A58125 | 1 kit |
The TaqPath Menu | GeneProof Treponema pallidum PCR Kit is a real-time PCR testing solution that is used to detect Treponema pallidum from swabs and urine samples. The kit is approved for use on the Applied Biosystems 7300/7500 real-time PCR systems (96-well).
Features of the TaqPath Menu | GeneProof Treponema pallidum PCR Kit include:
• High specific detection secured by highly conservative polA gene for DNA polymerase I
• Easily compatible with other GeneProof PCR kits in one workflow
• Easy to use—single master mix tube containing all components required for PCR amplification
• Minimizes contamination—the master mix contains uracil-DNA glycosylase (UNG) and dUTPs, reducing carryover contamination
• Compatible with a wide range of real-time PCR devices
The TaqPath Menu | GeneProof Treponema pallidum PCR Kit is a real-time PCR testing solution that is used for qualitative detection for diagnostics and aid to diagnostics of Treponema pallidum from swabs and urine samples. It is designed for professional use in laboratories with trained staff.