TrueMark™ Enteric Norovirus Select Amplification Control
Applied Biosystems™

TrueMark™ Enteric Norovirus Select Amplification Control

The TrueMark Enteric Norovirus Select Amplification Control contains a linearized multi-target plasmid for the following targets: norovirus GI, norovirus GII,Read more
Have Questions?
Catalog NumberQuantity
A5814125 μL
Catalog number A58141
Price (MXN)
-
Quantity:
25 μL

The TrueMark Enteric Norovirus Select Amplification Control contains a linearized multi-target plasmid for the following targets: norovirus GI, norovirus GII, and Bacillus atrophaeus.

This amplification control has a concentration of 1 x 105 copies/µL. It can be used as a positive control for TrueMark Enteric Norovirus Select Panel-specific amplification or as a stand-alone sample to verify assay performance and help with troubleshooting.

For Research Use Only.
Specifications
Quantity25 μL
Volume25 μL
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Real Time PCR (qPCR), RT-PCR
FormLiquid
PCR MethodMultiplex qPCR, qPCR, RT-PCR
Reaction Speed120 min.
Unit SizeEach