MagMAX™ Prime Viral/Pathogen Elution Buffer
Applied Biosystems™

MagMAX™ Prime Viral/Pathogen Elution Buffer

MagMAX Prime Viral/Pathogen Elution Buffer is offered separately here for applications that require more Elution Buffer than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Have Questions?
Catalog NumberQuantity
A58148120 mL
Catalog number A58148
Price (USD)
598.00
Each
Quantity:
120 mL
Price (USD)
598.00
Each

MagMAX Prime Viral/Pathogen Elution Buffer is offered separately here for applications that require more Elution Buffer than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.

For Research Use Only.

Specifications
Bead TypeMagnetic
ColorClear
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of ReactionsUp to 2000 Reactions
Nucleic Acid TypeDNA/RNA
Product LineMagMAX
Product TypeReagent
Quantity120 mL
Sample TypeSwabs in Transport Media, Saliva, Urine, Serum, Plasma
Sample Volume200–400 μL
Shelf Life2 Years
Shipping ConditionRoom Temperature
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
FormatBottle
Isolation TechnologyBeads
Unit SizeEach
Contents & Storage
Store at 15–25°C.