MagMAX™ Prime Viral/Pathogen Proteinase K Dye
Applied Biosystems™

MagMAX™ Prime Viral/Pathogen Proteinase K Dye

MagMAX Prime Viral/Pathogen Proteinase K Dye is offered separately here for applications that require more Proteinase K Dye than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.
Have Questions?
Catalog NumberQuantity
A58152150 μL
Catalog number A58152
Price (CLP)
62.778
Each
Quantity:
150 μL
Price (CLP)
62.778
Each

MagMAX Prime Viral/Pathogen Proteinase K Dye is offered separately here for applications that require more Proteinase K Dye than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.

For Research Use Only.
Specifications
Bead TypeMagnetic
ColorClear
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of ReactionsUp to 2000 Reactions
Nucleic Acid TypeDNA/RNA
Product LineMagMAX
Product TypeReagent
Quantity150 μL
Sample TypeSwabs in Transport Media, Saliva, Urine, Serum, Plasma
Sample Volume200–400 μL
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
FormatTube
Isolation TechnologyBeads
Unit SizeEach
Contents & Storage
Store at 15–25°C.