How can I tell if the buffer in which my peptides are resuspended are compatible with TMT and TMTpro labels?
TMT and TMTpro labels are not compatible with buffers containing primary amines. After protein digestion, peptides need to be cleaned up and dried down. Prior to labeling, we do recommend resuspending the peptides in 10 µL of either 100 mM TEAB (pH 8.5) or 100 mM HEPES (pH 8.0).
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.