TaqPath™ Menu ∣ GeneProof Factor XIII V34L PCR Kit
Applied Biosystems™

TaqPath™ Menu ∣ GeneProof Factor XIII V34L PCR Kit

The TaqPath Menu I GeneProof Factor XIII V34L PCR Kit is used for the qualitative detection of G103T polymorphism inRead more
Have Questions?
Catalog NumberQuantity
A58422100 reactions
Catalog number A58422
Price (EUR)
-
Quantity:
100 reactions

The TaqPath Menu I GeneProof Factor XIII V34L PCR Kit is used for the qualitative detection of G103T polymorphism in the F13A1 gene for Factor XIII (V34L) through real-time PCR technology. The kit is validated for use with whole blood specimens.

Features of the TaqPath Menu I GeneProof Factor XIII V34L PCR Kit include:
• Compatible with all other PCR kits from the thrombotic mutations panel in one workflow
• Easy to use—single ready-to-use master mix tube containing all components required for PCR amplification
• Simplifies laboratory workflow—no additional pipetting of PCR reagents
• Compatible with a wide range of real-time PCR devices

The kit is approved for use on Applied Biosystems 7300/7500 and QuantStudio 3/5 real-time PCR systems.

Access the Instructions for Use (IFU) approved for your country ›

Available in select countries only.
Specifications
Certification ComplianceCE-IVD
CompatibilityTested in Reference Institute for Bioanalytics External Quality Assessment Panels
For Use With (Equipment)QuantStudio™ 3/5 Real-Time PCR System, Applied Biosystems 7300/7500 StepOne™/StepOne Plus™ Real-Time PCR System, AriaMx Real-Time PCR System, CFX Connect™/CFX96™/Dx Real-Time PCR Detection System, LightCycler™ 2.0/480, LineGene 9600 Plus, Mic qPCR Cycler, croBEE™ Real-Time PCR System, Rotor-Gene™ 3000/6000/Q*, SLAN™ Real-Time PCR System
No. of Reactions100 reactions
Product LineTaqPath
Quantity100 reactions
Sample TypeWhole blood (EDTA)
SensitivityDiagnostic: 100% (CI95%: 84.50–100%)
TargetG103T Polymorphism in F13A1 gene for Factor XIII (V34L) (rs5985)
Target SpecificityDiagnostic: 100% (CI95%: 71.65–100%)
TechniqueQualitative analysis
Label or DyeFAM , HEX , JOE , VIC
PCR MethodqPCR
Unit SizeEach
Contents & Storage
• Master mix
• Positive control

Store at –20 ± 5°C.