TaqPath™ Menu ∣ GeneProof BK Virus (BKV) PCR Kit
Applied Biosystems

TaqPath™ Menu ∣ GeneProof BK Virus (BKV) PCR Kit

The TaqPath Menu I GeneProof BK Virus (BKV) PCR Kit is used for the qualitative and quantitative detection of BKMás información
Have Questions?
Número de catálogoCantidad
A584301 kit
Número de catálogo A58430
Precio (CLP)
1.458.464
Each
Cantidad:
1 kit
Precio (CLP)
1.458.464
Each

The TaqPath Menu I GeneProof BK Virus (BKV) PCR Kit is used for the qualitative and quantitative detection of BK virus through real-time PCR technology. It enables reliable viral load monitoring. The kit is validated for use with CSF, plasma, urine, and whole blood specimens.

Features of the TaqPath Menu I GeneProof BK Virus (BKV) PCR Kit include:
• Compatible with other PCR kits in one workflow
• Easy to use—single master mix tube containing all components required for PCR amplification
• Minimizes contamination—the master mix contains uracil-DNA glycosylase (UNG) and dUTPs, reducing carryover contamination
• Compatible with a wide range of real-time PCR devices

The kit is approved for use on Applied Biosystems 7300/7500 real-time PCR systems.

Access the Instructions for Use (IFU) approved for your country ›
Explore more transplant and immunocompromised infection solutions ›

Available in select countries only.
Especificaciones
Conformidad de la certificaciónCE-IVD
CompatibilidadRegularly tested in QCMD and INSTAND e.V. External Quality Assessment Panels
Molde controlPCR inhibition and DNA extraction efficiency control by Internal Control (IC)
Para utilizar con (equipo)Applied Biosystems 7300/7500 Real-Time PCR System, croBEE Real-Time PCR System, AMPLilab Real-Time PCR System, AriaMx Real-Time PCR System, BioQuant-96 Real-Time PCR System, CFX Connect™/CFX96™/Dx Real-Time PCR Detection System, Gentier 96E/96R Real-Time PCR System, LineGene 9600 Plus, Mic qPCR Cycler, Rotor-Gene 3000/Q, SLAN™ Real-Time PCR System
N.º de reacciones100 reacciones
Línea de productosTaqPath
Cantidad1 kit
Tipo de muestraCSF, plasma, urine, whole blood
SensibilidadDiagnostic: 96.30% (CI95%: 79.11–99.81%)
ObjetivoT-Ag gene encoding large tumor antigen
Especificidad de dianaDiagnostic: 100.00% (CI95%: 86.27–100.00%)
TécnicaQualitative and Quantitative Analysis
Etiqueta o tinteFAM, HEX, JOE
Método de PCRqPCR, RT-PCR
Unit SizeEach
Contenido y almacenamiento
• Mastermix
• Calibrators
• Internal Control

Store at –20 ± 5°C.