TaqPath™ Menu ∣ GeneProof™ BK/JC Virus (BK/JC) PCR Kit
Applied Biosystems™

TaqPath™ Menu ∣ GeneProof™ BK/JC Virus (BK/JC) PCR Kit

The TaqPath Menu I GeneProof BK/JC Virus (BK/JC) PCR Kit is used for the qualitative and quantitative detection of BKMás información
Have Questions?
Número de catálogoCantidad
A584311 kit
Número de catálogo A58431
Precio (CLP)
1.662.192
Each
Cantidad:
1 kit
Precio (CLP)
1.662.192
Each

The TaqPath Menu I GeneProof BK/JC Virus (BK/JC) PCR Kit is used for the qualitative and quantitative detection of BK and JC virus through real-time PCR technology. The kit detects and distinguishes Bordetella pertussis and Bordetella parapertussis in a single reaction tube. The kit is validated for use with CSF, plasma, urine, and whole blood specimens.

Features of the TaqPath Menu I GeneProof BK/JC Virus (BK/JC) PCR Kit include:
• Compatible with other PCR kits in one workflow
• Easy to use—single master mix tube containing all components required for PCR amplification
• Minimizes contamination—the master mix contains uracil-DNA glycosylase (UNG) and dUTPs, reducing carryover contamination
• Compatible with a wide range of real-time PCR devices

The kit is approved for use on Applied Biosystems 7500 and QuantStudio 5 real-time PCR systems.

Access the Instructions for Use (IFU) approved for your country ›
Explore more transplant and Immunocompromised infection solutions ›

Available in select countries only.
Especificaciones
Conformidad de la certificaciónCE IVD
CompatibilidadRegularly tested in QCMD and INSTAND e.V. External Quality Assessment Panels
Molde controlPCR inhibition and DNA extraction efficiency control by Internal Control
Para utilizar con (equipo)QuantStudio™ 5 Real-Time PCR System, Applied Biosystems 7500 Real-Time PCR System, croBEE Real-Time PCR System, AMPLilab Real-Time PCR System, AriaMx Real-Time PCR System, BioQuant-96 Real-Time PCR System, CFX96™/Dx Real-Time PCR Detection System, Gentier 96E/96R Real-Time PCR System, LightCycler™ 480, LineGene 9600 Plus, Rotor-Gene 3000/Q, SLAN™ Real-Time PCR System
N.º de reacciones100 reacciones
Línea de productosTaqPath
Cantidad1 kit
Tipo de muestraCSF, plasma, urine, whole blood
SensibilidadDiagnostic: 100.00%
ObjetivoBK Virus (BKV) and JC Virus (JCV) (T-Ag gene encoding large tumor antigen (BKV) and t-Ag gene encoding small tumor antigen (JCV))
Especificidad de dianaAnalytical: 100% (BK Virus), 100% (JC Virus)
Diagnostic: 100.00%
TécnicaQualitative and Quantitative Analysis
Etiqueta o tinteFAM, Cy5, HEX, JOE, VIC
Método de PCRqPCR, RT-PCR
Unit SizeEach
Contenido y almacenamiento
• Mastermix
• Calibrators
• Internal Control

Store at –20 ± 5°C.