TaqPath™ Menu ∣ GeneProof Varicella-Zoster Virus (VZV) PCR Kit
Applied Biosystems

TaqPath™ Menu ∣ GeneProof Varicella-Zoster Virus (VZV) PCR Kit

The TaqPath Menu I GeneProof Varicella-Zoster Virus (VZV) PCR Kit is used for the qualitative and quantitative detection of Varicella-ZosterRead more
Have Questions?
Catalog NumberQuantity
A58432100 reactions kit
Catalog number A58432
Price (EUR)
-
Quantity:
100 reactions kit

The TaqPath Menu I GeneProof Varicella-Zoster Virus (VZV) PCR Kit is used for the qualitative and quantitative detection of Varicella-Zoster virus through real-time PCR technology. The kit is validated for use with CSF, plasma, serum, whole blood (EDTA), and vesicular swab specimens.

Features of the TaqPath Menu I GeneProof Varicella-Zoster Virus (VZV) PCR Kit include:
• Compatible with other PCR kits in one workflow
• Easy to use—single master mix tube containing all components required for PCR amplification
• Minimizes contamination—the master mix contains uracil-DNA glycosylase (UNG) and dUTPs, reducing carryover contamination
• Compatible with a wide range of real-time PCR devices

The kit is approved for use on Applied Biosystems 7300/7500 and QuantStudio 3/5 real-time PCR systems.

Access the Instructions for Use (IFU) approved for your country ›
Explore more transplant and immunocompromised infection solutions ›

Available in select countries only.
Specifications
Certification ComplianceCE-IVD
CompatibilityRegularly tested in QCMD and INSTAND e.V. External Quality Assessment Panels
Control TemplatePCR inhibition and DNA extraction efficiency control by Internal Control (IC)
For Use With (Equipment)QuantStudio™ 3/5 Real-Time PCR System, Applied Biosystems 7300/7500 Real-Time PCR System, StepOne™/StepOne Plus™ Real-Time PCR System, croBEE Real-Time PCR System, AMPLilab Real-Time PCR System, AriaMx Real-Time PCR System, BioQuant-96 Real-Time PCR System, CFX Connect™/CFX96™/Dx Real-Time PCR Detection System, Gentier 96E/96R Real-Time PCR System, LightCycler™ 2.0/480, LineGene 9600 Plus, Mic qPCR Cycler, Rotor-Gene 3000/Q, SLAN™ Real-Time PCR System
No. of Reactions100 Reactions
Product LineTaqPath
Quantity100 reactions kit
Sample TypeCSF, plasma, serum, whole blood (EDTA), vesicular swab (For qualtitative evaluation only)
SensitivityDiagnostic: 100.00% (CI95%: 97.52–100.00%)
TargetORF31 gene
Target SpecificityDiagnostic: 97.44% (CI95%: 84.92–99.87%)
TechniqueQualitative and Quantitative Analysis
Label or DyeFAM, HEX, JOE, VIC
PCR MethodqPCR, RT-qPCR
Unit SizeEach
Contents & Storage
• Mastermix
• Calibrators
• Internal Control

Store at –20 ± 5°C.