Penicillin Streptomycin GlutaMAX™ Supplement (100X) - FAQs
Penicillin Streptomycin GlutaMAX™ Supplement (100X) - FAQs
View additional product information for Penicillin Streptomycin GlutaMAX™ Supplement (100X) - FAQs (A5873601)
12 product FAQs found
What is the concentration of each component in Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601)?
Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) contains the antibiotics penicillin (10,000 units) and streptomycin (10,000 µg), along with 200 mM L-alanyl-L-glutamine dipeptide, GlutaMAX supplement, in a citrate buffer to maintain penicillin potency.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the difference between Penicillin-Streptomycin-Glutamine (100X) (Cat. No. 10378016) and Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601)?
Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) is an alternative to Penicillin-Streptomycin-Glutamine (100X) (Cat. No. 10378016), with increased stability that improves cell health. In aqueous solutions such as cell culture media, L-glutamine spontaneously degrades, generating toxic ammonia and pyrrolidine carboxylic acid as byproducts. GlutaMAX supplement is a dipeptide, L-alanyl-L-glutamine, which is more stable in cell culture media.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
When I received Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601), the supplement was partially thawed in the bottle. Will it still work?
Yes, the product will still work. The bottle can be stored at 4 degrees C overnight to finish thawing and then aliquoted the next day for storage at -20 degrees C. Alternatively, the bottle can be stored at -20 degrees C until it is needed.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Our -20 degrees C storage space is full. Can I store Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) at -80 degrees C instead?
Yes, Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) may be stored at -80 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How often should medium changes be performed for cells cultured in medium prepared with Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601)?
Medium changes should be performed at least once every three days as penicillin and streptomycin are stable for three days at 37 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How long is Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) stable in solution?
Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) is stable for two weeks in prepared cell culture media stored at 4 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Can I use Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) in combination with other antibiotic reagents?
This would need to be empirically determined for each cell line testing each combination with other antibiotics. Penicillin and streptomycin are effective against gram-negative and gram-positive bacteria.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
Will Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) affect the expansion rate of cells?
When Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) is diluted to 1X, it is designed to have equivalent performance to 1X dilutions of Penicillin-Streptomycin (100X) (Cat. No. 15140122) and GlutaMAX Supplement (100X) (Cat. No. 35050061). Cells previously maintained in medium containing 1X dilutions of Penicillin-Streptomycin (100X) and GlutaMAX Supplement (100X) should have the same rate of expansion when transitioned to Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the shelf life of Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601)?
The shelf life of Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) is 12 months.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How should I store Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601)?
We recommend storing Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) at -20 degrees C until the first thaw. Upon thawing the entire contents of the bottle, the supplement can be aliquoted at appropriate volumes into small tubes for convenient future use and the aliquots can be stored at -20 degrees C. It is best to aliquot what is required for single-use experiments prior to storage at -20 degrees C.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
How should I thaw Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601)?
For convenience, we recommend thawing the entire bottle of Penicillin Streptomycin GlutaMAX Supplement (100X) at 4 degrees C overnight.
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.
What is the proper dilution to use for Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601)?
Penicillin Streptomycin GlutaMAX Supplement (100X) (Cat. No. A5873601) is intended to be used at a 1:100 dilution as a direct replacement for Penicillin-Streptomycin (100X) (Cat. No. 15140122) and GlutaMAX Supplement (100X) (Cat. No. 35050061). Penicillin Streptomycin GlutaMAX Supplement (100X) is also an alternative for Penicillin-Streptomycin-Glutamine (100X) (Cat. No. 10378016).
Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.