MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer
Applied Biosystems™

MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호반응 수
A59053Up to 100 Reactions
A58153Up to 1000 Reactions
카탈로그 번호 A59053
제품 가격(KRW)
357,000
온라인 행사
Ends: 31-Dec-2026
420,000
할인액 63,000 (15%)
Each
반응 수:
Up to 100 Reactions
제품 가격(KRW)
357,000
온라인 행사
Ends: 31-Dec-2026
420,000
할인액 63,000 (15%)
Each

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi. It is designed to be used in conjunction with MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.

For Research Use Only.
사양
비드 유형Magnetic
색상Clear
용도(애플리케이션)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
고처리량 호환성High-throughput Compatible, Automated Protocols
주요 기능Isolate and purify viral, bacterial, parasitic, and fungal nucleic acid
반응 수Up to 100 Reactions
핵산 유형DNA/RNA
제품라인MagMAX
제품 유형Reagent
수량5 mL
샘플 종류Swabs in Transport Media, Saliva, Urine, Serum, Plasma
샘플 부피200–400 μL
배송 조건Dry Ice
타겟Viral, Bacterial, Parasitic, and Fungal Nucleic Acid
형식Bottle
Isolation TechnologyBeads
Unit SizeEach
구성 및 보관
Store at –15 to –25°C.