Tris 20 mM, NaCl 1.5 M
Gibco™
Tris 20 mM, NaCl 1.5 M
Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.
製品番号(カタログ番号)数量
A59168BE200 L
A59168BA5 L
A59168BB20 L
A59168BC50 L
A59168BD100 L
製品番号(カタログ番号) A59168BE
価格(JPY)
-
数量:
200 L
Gibco process buffers are sterile-filtered, packaged in bioprocess containers, and ready to order. Sourcing process buffers can allow you to focus on additional critical steps in your biopharmaceutical processes.
Features
  • Available in a range of pack sizes
  • Established shelf life; set specifications
  • Made to order
  • Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
  • Readily accessible documentation to help accelerate qualification
Benefits
  • Helps ease time requirements for in-house buffer preparation
  • Mitigates space constraints for equipment storage
  • Streamlines in-house processes related to formulation, hydration, testing, and validation
  • Addresses workflow needs for biologics development and manufacturing stages
Additional information
  • Multi-compendial testing: compendial-grade or equivalent raw materials
  • Primary packaging: Aegis 5-14 single-use bioprocess container (BPC)
  • Secondary packaging: rigid shipping container with appropriate dunnage
  • Manufacturing facilities: ISO 13485–certified, cGMP-compliant to 21 CFR Part 820 standard
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
仕様
組成Tris/NaCl Buffer
製造品質CGMP
数量200 L
形状Clear, colorless solution
製品タイプSodium Chloride
無菌性Sterile-filtered
Sterilization MethodSterile-filtered
Unit SizeEach