Live Cell Imaging Solution
Invitrogen™

Live Cell Imaging Solution

The Live Cell Imaging Solution from Molecular Probes™ is a physiological medium developed for live cell imaging applications that hasLeia mais
Have Questions?
Número do catálogoQuantity
A59688DJ500 mL
Número do catálogo A59688DJ
Preço (BRL)
260,17
Each
Quantity:
500 mL
Preço (BRL)
260,17
Each

The Live Cell Imaging Solution from Molecular Probes™ is a physiological medium developed for live cell imaging applications that has universal utility in cell-based research—for live cell imaging, dye loading, and wash steps. It is an optically clear, physiological solution buffered with HEPES at pH 7.4 that keeps cells healthy for up to 4 hours at ambient atmosphere and temperature. Compared to standard media, Live Cell Imaging Solution provides better clarity, signal-to-noise ratio, and cell viability.

Outstanding Features of Live Cell Imaging Solution:
• Physiological and chemical optimization—providing the ideal solution for live cell fluorescence imaging applications
• Optical clarity—affording more efficient imaging and minimal background
• Extended buffering capacity compared to media—for improved cell viability

Obtain Great Results with a Medium Designed to Meet the Demands of Live Cell Imaging
Live cell imaging can be challenging. Not only must staining be optimized for the particular assay readout, spectral compatibility, and signal-to-noise level, but live cell imaging of dynamic processes and cell physiology requires live observation over time. The choice of medium is particularly important for time-lapse imaging and experiments where cells are exposed to ambient conditions for longer periods. For reliable results, it is essential that cells be healthy and maintained as closely as possible to physiological temperature, pH, oxygen tension, and other conditions. If cells appear unhealthy they should not be used for imaging. Live Cell Imaging Solution has been developed specifically for live cell imaging applications; as a result, it provides image clarity (Fig 1), signal-to-noise ratio (Fig 2), and cell viability that are significantly enhanced compared to standard media. In addition to live cell imaging, the physiological composition of the Live Cell Imaging Solution makes it ideal for other cell-based applications, such as wash steps and dye loading.

Need help deciding what product you need for your cell imaging? See our cell imaging workflow and decision tree.

Tired of poor signal-to-noise? See our collection antifade and signal enhancer products from Molecular Probes™.

For Research Use Only.
Especificações
Quantity500 mL
Reagent TypePhysiological Imaging Buffer
Shipping ConditionRoom Temperature
Product LineMolecular Probes
Unit SizeEach
Conteúdo e armazenamento

Store at ≤25°C.

Frequently asked questions (FAQs)

What is the formulation of the Live Cell Imaging Solution (Cat. No. A59688DJ)?

The formulation of the Live Cell Imaging Solution (Cat. No. A59688DJ) is shown below:
140 mM NaCl
2.5 mM KCl
1.8 mM CaCl2
1.0 mM MgCl2
20 mM HEPES
pH 7.4
mOsm = 300

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.