VetMAX™ African Swine Fever Virus MGF360-CD2V DIVA Reagents
Applied Biosystems™

VetMAX™ African Swine Fever Virus MGF360-CD2V DIVA Reagents

The VetMAX African Swine Fever Virus MGF360-CD2V DIVA Reagents kit contains a single tube with the primer/probes and master mix needed for a qPCR assay targeting the CD2V and MGF360-14L genes of African swine fever virus.
Have Questions?
Catalog NumberQuantity
A59802500 Reactions
Catalog number A59802
Price (CLP)
-
Quantity:
500 Reactions

The VetMAX African Swine Fever Virus MGF360-CD2V DIVA Reagents kit contains a single tube with the primer/probes and master mix needed for a qPCR assay targeting the CD2V and MGF360-14L genes of African swine fever virus. It offers rapid testing and can be used as reflex assay to investigate samples from animals vaccinated with CD2V and MGF360-14L recombinant vaccine after screening for African Swine Fever with the VetMAX African Swine Fever Virus Detection Kit (Cat. No. A28809). Positive controls for the assay are available separately (Cat. No. A59803).

Features of the VetMAX African Swine Fever Virus MGF360-CD2V DIVA Reagents include:
• Ready-to-use master mix
• Specific design for CD2V and MGF360-14L recombinant vaccine
• Express assay run time (∼1 hour)
• Can be run simultaneously with the VetMAX African Swine Fever Virus Detection Kit

African swine fever is a highly contagious viral disease of pigs and wild boars. African swine fever virus outbreaks spread rapidly in pig populations through direct contact and the bite of infected soft tick species. Outbreaks cause commerce disruption resulting in severe economic losses in pig-producing countries. Newly approved vaccines are now deployed in some pig production countries. Rapid differentiation of infected vs vaccinated animals (DIVA) by PCR is fundamental to control the disease.

For Research Use Only.
Specifications
DiseaseAfrican Swine Fever
For Use With (Equipment)QuantStudio™ 5, 7500 Fast Thermocycler
High-throughput CompatibilityYes
No. of Reactions500 reactions
Passive Reference DyeROX
Quantity500 Reactions
Shelf Life12 Months
Shipping ConditionFrozen
SpeciesSwine
Volume10 μL
Detection MethodqPCR
For Use With (Application)Pathogen Detection, Virus Detection
FormLiquid
GC-Rich PCR PerformanceMedium
Label or Dye6-FAM
PCR MethodqPCR
Reaction Speed15 to 30 min.
Unit Size500 reactions