CMAC, <i>t</i>-BOC-Leu-Met (7-Amino-4-Chloromethylcoumarin, <i>t</i>-BOC-L-Leucyl-L-Methionine amide)
Invitrogen™

CMAC, t-BOC-Leu-Met (7-Amino-4-Chloromethylcoumarin, t-BOC-L-Leucyl-L-Methionine amide)

El sustrato de peptidasa de CMAC, t-BOC-Leu-MET, puede usarse para medir la actividad de la peptidasa en solución o enMás información
Have Questions?
Número de catálogoCantidad
A65205 mg
Número de catálogo A6520
Precio (MXN)
-
Cantidad:
5 mg
El sustrato de peptidasa de CMAC, t-BOC-Leu-MET, puede usarse para medir la actividad de la peptidasa en solución o en células vivas. El sustrato tiene un máximo de excitación/emisión de ∼ 330/403. Después de la escisión por peptidasas, el producto produce fluorescencia azul con un máximo de excitación/emisión de ∼ 351/430 nm.
Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Permeabilidad celularPermeabilidad celular
Cantidad5 mg
Condiciones de envíoTemperatura ambiente
SustratoSustrato de peptidasa de CMAC
Método de detecciónFluorescente
FormularioSólido
Substrate PropertiesSustrato basado en péptidos
Tipo de sustratoSustrato de peptidasa
Target EnzymeCalpaína
Unit SizeEach
Contenido y almacenamiento
Almacenar en congelador (de -5 a -30 °C).

Citations & References (20)

Citations & References
Abstract
Calpain-mediated X-linked inhibitor of apoptosis degradation in neutrophil apoptosis and its impairment in chronic neutrophilic leukemia.
Authors: Kobayashi Susumu; Yamashita Kouhei; Takeoka Tomoharu; Ohtsuki Tetsuya; Suzuki Yasuyuki; Takahashi Ryosuke; Yamamoto Kokichi; Kaufmann Scott H; Uchiyama Takashi; Sasada Masataka; Takahashi Atsushi;
Journal:J Biol Chem
PubMed ID:12121983
'The number of neutrophils in the blood and tissues is controlled by constitutive apoptotic programmed cell death and clearance by phagocytes such as macrophages. Here, we found that calpains cleave the X-linked inhibitor of apoptosis (XIAP) in vitro, producing fragments that are unable to inhibit caspase-3. These fragments were detected ... More
Suppression of cancer cell migration and invasion by protein phosphatase 2A through dephosphorylation of mu- and m-calpains.
Authors:Xu L, Deng X
Journal:J Biol Chem
PubMed ID:16982626
'The mu- and m-calpains are major members of the calpain family that play an essential role in regulating cell motility. We have recently discovered that nicotine-activated protein kinase C iota enhances calpain phosphorylation in association with enhanced calpain activity and accelerated migration and invasion of human lung cancer cells. Here ... More
Protein kinase Ciota promotes nicotine-induced migration and invasion of cancer cells via phosphorylation of micro- and m-calpains.
Authors:Xu L, Deng X
Journal:J Biol Chem
PubMed ID:16361262
'Nicotine is a major component in cigarette smoke that activates the growth-promoting pathways to facilitate the development of lung cancer. However, it is not clear whether nicotine affects cell motility to facilitate tumor metastasis. Here we discovered that nicotine potently induces phosphorylation of both mu- and m-calpains via activation of ... More
Tumor necrosis factor-alpha-inducible IkappaBalpha proteolysis mediated by cytosolic m-calpain. A mechanism parallel to the ubiquitin-proteasome pathway for nuclear factor-kappab activation.
Authors:Han Y, Weinman S, Boldogh I, Walker RK, Brasier AR
Journal:J Biol Chem
PubMed ID:9873017
'The cytokine tumor necrosis factor alpha (TNF-alpha) induces expression of inflammatory gene networks by activating cytoplasmic to nuclear translocation of the nuclear factor-kappaB (NF-kappaB) transcription factor. NF-kappaB activation results from sequential phosphorylation and hydrolysis of the cytoplasmic inhibitor, IkappaBalpha, through the 26 S proteasome. Here, we show a parallel proteasome-independent ... More
Calpain is required for normal osteoclast function and is down-regulated by calcitonin.
Authors:Marzia M, Chiusaroli R, Neff L, Kim NY, Chishti AH, Baron R, Horne WC
Journal:J Biol Chem
PubMed ID:16461769
'Osteoclast motility is thought to depend on rapid podosome assembly and disassembly. Both mu-calpain and m-calpain, which promote the formation and disassembly of focal adhesions, were observed in the podosome belt of osteoclasts. Calpain inhibitors disrupted the podosome belt, blocked the constitutive cleavage of the calpain substrates filamin A, talin, ... More