TrueMark™ PGx Comprehensive Plasmid Controls
Applied Biosystems™

TrueMark™ PGx Comprehensive Plasmid Controls

TrueMark PGx Comprehensive Plasmid Controls contains three linearized multi-target plasmid pools with human genome target sequences commonly used for pharmacogenomic research testing.
Have Questions?
Catalog NumberQuantity
A653421 Kit, 15 Tubes total
Catalog number A65342
Price (USD)
2,435.00
Each
Quantity:
1 Kit, 15 Tubes total
Price (USD)
2,435.00
Each

TrueMark PGx Comprehensive Plasmid Controls contains three linearized multi-target plasmid pool controls with human genome target sequences commonly used for pharmacogenomics research testing.

These controls were designed for use with TaqMan real-time qPCR assays to verify the presence of homozygous and heterozygous genotyping, copy number variation (CNV), and CNV reference assays (TERT and RNase P). This control kit provides coverage of essential, commonly studied markers within CYP2D6, CYP2C19, CYP2C9, DPYD, and other important DME and CNV research genes.

The three controls are supplied at a concentration of ~1 x 105 copies/μL in 5 tubes of 50 μL each. They are quantified using digital PCR for accuracy and can be used as stand-alone samples to assess assay performance or to assist with assay troubleshooting. They may also serve as a CNV positive run control/calibrator.

Compatible OpenArray and TaqMan array plates/panels

  • TaqMan OpenArray PGx Express 60 Panel, Cat. No. 4488847
  • TaqMan PGx Express Panel (64), 384-well Plate, Cat. No. A37610
  • TaqMan OpenArray PGx Express 120 Panel (v3)
  • TrueMark PGx SNP + CNV, 64 Panel, 384-well plate, Cat. No. A40003215
  • TrueMark PGx SNP + CNV, 64 Panel, OpenArray
  • TrueMark PGx SNP + CNV, 128 Panel, OpenArray

If your assay is not supported with this product, please consider the TaqMan Custom DNA Controls offered by GeneArt. This product is manufactured in an ISO 9001 compliant facility.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
No. of Reactions3 control pools (5 aliquiots x 50 μL each pool)
Quantity1 Kit, 15 Tubes total
Shelf Life24 Months
Shipping ConditionDry Ice
TechniqueReal-time qPCR
Volume50 μL
Concentration1 x 105 Copies/μL
Detection MethodTaqMan
For Use With (Application)Genotyping, Copy Number Variation Analysis, Real Time PCR (qPCR), Digital PCR (dPCR)
FormFrozen
PCR MethodqPCR, Digital PCR
Unit SizeEach