SuperScript™ IV Template Switching RT Master Mix
Invitrogen™

SuperScript™ IV Template Switching RT Master Mix

Invitrogen SuperScript IV Template-Switching RT Master Mix is designed for high template switching efficiency in cDNA synthesis reactions. It containsRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A6542496 Reactions
A6542348 Reactions
A654255 x 96 Reactions
Catalog number A65424
Price (USD)
1,116.00
Each
No. of Reactions:
96 Reactions
Price (USD)
1,116.00
Each

Invitrogen SuperScript IV Template-Switching RT Master Mix is designed for high template switching efficiency in cDNA synthesis reactions. It contains SuperScript IV Reverse Transcriptase, RNase Inhibitor, and all other reverse transcription reaction components in a convenient premixed format. Only RNA template, RT primer, and template switching oligo (TSO) of choice need to be added.

Features of SuperScript IV Template-Switching RT Master Mix include:
• Formulated for high quality full-length cDNA synthesis
• Includes proprietary enhancer to maximize template switching efficiency
• Exceptional sensitivity for easy detection of low abundance targets (as low as single cell or 2 pg total RNA)
• Cell lysis buffer included for direct cDNA synthesis without RNA purification

Procedure overview
To maximize template switching efficiency, SuperScript IV Template-Switching RT Master Mix contains proprietary template switching enhancer and leverages the terminal deoxynucleotidyl transferase (TdT) activity of SuperScript IV Reverse Transcriptase. When the reverse transcriptase reaches the 5’ end of the RNA template, TdT activity adds 1–3 extra nucleotides to the cDNA end, enabling binding of the template switching oligo and incorporation of known sequence at the 3’ end of the resulting cDNA. This cDNA can be used for amplification by PCR, gene expression analysis by real-time PCR, or as a template for 5’RACE, RNA-seq, or second strand cDNA synthesis.This master mix is compatible with a wide input range of total RNA (from 2 pg to 4 µg) and is supplied with lysis buffer to enable direct cDNA synthesis from mammalian cells (1–1,000 cells per sample).

Related products
SuperScript IV Template-Switching RT Master Mix is compatible with various RT primers and self-designed template switching oligos. We offer Template Switching Oligo and Capturing Oligo (dT) Primer, both containing the same adaptor sequence complementary to Preamplification Primer. Alternatively, a full set of reagents for template switching cDNA synthesis and preamplification is available in the SuperScript IV Single Cell/Low-Input cDNA PreAmp Kit.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Final Product TypeFirst-Strand cDNA
FormatMaster Mix
No. of Reactions96 Reactions
Quantity96 reactions
Reagent TypeReverse Transcription
Starting MaterialCells or RNA
TechniqueTemplate Switching Reverse Transcription
Concentration4X
GC-Rich PCR PerformanceHigh
Reaction Speed30 min.
Unit SizeEach
Contents & Storage
• 4X SuperScript IV cDNA Synthesis Master Mix (480 μL)
• 10X Lysis Buffer (96 μL)
• Nuclease-free water (1.25 mL)

Store at -15 to -25°C.

Frequently asked questions (FAQs)

What template switching oligos can be used with SuperScript IV Template Switching Master Mix?

Invitrogen Template Switching Oligo (Cat. No. A65427) is offered, in conjunction with Capturing Oligo (dT) (Cat. No. A65429) and Preamplification Primers (Cat. No. A65428), complementary to the TSO. Alternatively, custom self-designed template switching oligo can be used.

Can SuperScript IV Template Switching RT Master Mix be used for other applications that do not involve template switching, e.g. RT-qPCR?

Yes, the master mix can be used for the first strand cDNA synthesis without templates switching oligo, but the results may be not as good as with more specialized product. For RT-qPCR, we recommend SuperScript IV VILO Master Mix.

SuperScript IV Template Swiching RT Master Mix is supplied with lysis buffer. Why does the protocol recommend using lysis buffer even with purified RNA samples?

Lysis buffer supplied with SuperScript IV Template Switching RT Master Mix has a dual function. It enables direct cDNA synthesis workflow without RNA purification, and it includes additional amount of proprietary enhancer which improves templates switching efficiency with low input samples of purified RNA (up to 10 ng).

SuperScript IV Template Switching RT Master Mix is supplied with lysis buffer. Why does the protocol recommend using lysis buffer even with purified RNA samples?

Lysis buffer supplied with SuperScript IV Template Switching RT Master Mix has a dual function. It enables direct cDNA synthesis workflow without RNA purification, and it includes an additional amount of proprietary enhancer which improves templates switching efficiency with low input samples of purified RNA (up to 10 ng).