POROS™ CaptureSelect™ C1-InhXL Affinity Resin
Thermo Scientific™

POROS™ CaptureSelect™ C1-InhXL Affinity Resin

Thermo Scientific™ POROS™ CaptureSelect™ C1-InhXL Affinity Resin is intended for the purification of human C1 esterase inhibitor from recombinant and plasma/serum sources.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
A65439250 mL
A6543625 mL
A6543750 mL
A654401 L
A654415 L
A6544210 L
Número de catálogo A65439
Precio (MXN)
-
Cantidad:
250 mL

POROS CaptureSelect C1-InhXL Affinity Resin is intended for the purification of human C1 esterase inhibitor from recombinant and plasma/serum sources. C1 esterase inhibitor is a protease inhibitor belonging to the serpin superfamily that inhibits the spontaneous activation of the complement system. The CaptureSelect C1-InhXL affinity ligand is highly selective for human C1 inhibitor and coupled to the high-performing POROS resin, allowing for an efficient and scalable purification process, with high purity and yield obtained in a single step. The large pore structure of the POROS beads enables higher flowrates and efficient binding of larger biomolecules.

Features of POROS CaptureSelect C1-InhXL Affinity Resin include:

  • Efficient purification of C1 esterase inhibitor with high yield
  • Mild pH elution condition to retain activity
  • High binding capacity at high flow rates
  • Excellent scalability
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics

  • Resin: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Average particle size: 50 μm
  • Ligand: single-domain [VHH] antibody fragment
  • Binding capacity: 20–25 g C1-Inhibitor/L resin
  • Elution conditions: 50 mM sodium acetate, 1.0 M MgCl2, pH 5.0
  • Regeneration conditions: acidic stripping condition with 0.5 M acetic acid, pH 2
  • Mechanical resistance: 100 bar (1450 psi, 10 MPa)
  • Formulation buffer: 18% (v/v) ethanol
For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to humans or animals.
Especificaciones
Capacidad de unión20 to 25 g C1-Inhibitor/L Resin
Para utilizar con (aplicación)Protein purification
Cantidad250 mL
Condiciones de envíoWet Ice
ObjetivoC1 Esterase Inhibitor
FormularioPulpa de resina
Tamaño de partículas50 μm
Línea de productosPOROS CaptureSelect
TipoAffinity Resin
Unit SizeEach
Contenido y almacenamiento
Store at 2°C to 8°C.