TaqMan™ Cells-to-CT™ HepatoExpress™ Kit
Invitrogen™

TaqMan™ Cells-to-CT™ HepatoExpress™ Kit

The TaqMan Cells-to-CT HepatoExpress Kit is a complete cell lysate system designed for gene expression analysis directly from hepatocytes without the need of RNA purification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A6578840 Reactions
A65789100 Reactions
A65790400 Reactions
A657912500 Reactions (Lysis only)
Catalog number A65788
Price (USD)
1,045.00
Each
No. of Reactions:
40 Reactions
Price (USD)
1,045.00
Each

The TaqMan™ Cells-to-CT™ HepatoExpress Kit is a hepatocyte gene expression kit designed for RNA extraction-free RT-qPCR. It enables fast, direct lysis of primary hepatocytes, eliminating the need for RNA purification while reducing sample preparation time to ~10 minutes.

This streamlined cell-to-CT workflow allows researchers to perform high-sensitivity gene expression analysis directly from cells, without columns, beads, or centrifugation. The result is reliable, reproducible RT-qPCR data with significantly reduced hands-on time.

Optimized specifically for primary hepatocyte models—including ECM-overlay cultures—the kit supports high-throughput, automation-compatible workflows for drug metabolism, toxicity studies, and mechanistic research.

Key Benefits

Fast, Extraction-Free Gene Expression Workflow

  • Perform RT-qPCR without RNA isolation or purification
  • Reduce RNA preparation time from hours to ~10 minutes
  • Direct lysis workflow minimizes sample handling and variability

High-Quality RT-qPCR Performance

  • Sensitivity and reproducibility comparable to purified RNA workflows
  • Preserves RNA integrity during lysis
  • Compatible with multiplex qPCR and low-input samples

Optimized for Primary Hepatocytes

  • Efficient lysis of hepatocytes, including ECM-overlay cultures
  • Consistent performance across hepatocyte species and models
  • Designed for challenging in vitro liver systems

Automation-Ready and Scalable

  • Compatible with 96-well plate formats and robotic liquid handling systems
  • An ideal choice for high-throughput screening and large-scale studies
  • Room-temperature workflow simplifies lab integration

Sustainable and Simplified Workflow

  • Significantly reduces plastic consumables associated with RNA purification
  • REACH-compliant reagents support environmentally responsible lab practices

Designed for Hepatocyte Gene Expression Challenges

Primary hepatocytes are widely used for drug metabolism, CYP450 induction, and toxicity studies, but efficient RNA extraction from these cells—especially in ECM-overlay cultures—can be difficult and time-consuming.

The HepatoExpress Kit addresses these challenges with a direct lysis chemistry that:

  • Effectively penetrates extracellular matrix (ECM) overlays
  • Protects RNA during lysis without purification
  • Maintains compatibility with reverse transcription and qPCR

This enables accurate gene expression analysis without the variability introduced by traditional RNA extraction workflows.

Applications

The HepatoExpress Kit is an ideal choice for:

  • Hepatocyte gene expression analysis
  • CYP450 induction assays
  • Drug metabolism and ADME studies
  • Toxicology and safety screening
  • Target verification and early-stage compound screening
  • Mechanistic studies in liver biology

Scalable from Discovery to High-Throughput Screening

Whether performing small-scale mechanistic studies or large-scale screening, the kit supports flexible experimental design. Its automation-friendly format enables seamless integration into high-throughput workflows, improving reproducibility and throughput.

Workflow: From Cells to qPCR in ~75 Minutes

The simple three-step workflow enables rapid gene expression analysis:

  1. Cell lysis (~10 minutes, room temperature)
    • Simultaneous cell lysis, RNA preservation, and genomic DNA removal
  2. Reverse transcription
    • cDNA synthesis using SuperScript™ IV VILO™ Master Mix
  3. qPCR detection
    • Sensitive, multiplex-capable amplification using TaqMan™ Gene Expression Assays

Compatible with 700,000+ TaqMan™ Gene Expression Assays, enabling broad pathway and target coverage.

Why Choose an RNA Extraction-Free qPCR Workflow?

Compared to traditional column-based RNA isolation methods, the HepatoExpress Kit:

  • Reduces workflow time from hours to minutes
  • Minimizes sample loss and variability
  • Reduces multiple handling steps that can degrade RNA
  • Improves consistency across replicates and experiments

The TaqMan™ Cells-to-CT™ HepatoExpress Kit is a fast, reliable, and scalable solution for primary hepatocyte gene expression analysis. By enabling direct, RNA extraction-free RT-qPCR, it simplifies workflows while maintaining high data quality—making it excellent for modern drug discovery and in vitro liver research.

Addition to HepatoExpress Product Description:

In addition to the applications listed above, the HepatoExpress Cells-to-CT Kit may be suitable for use with 3D culture systems based on customer-reported experience across a range of experimental models. These reports suggest that the kit can be incorporated into workflows involving complex cellular architectures, such as spheroids or organoid cultures, without the need for extensive protocol modification. However, this application has not been independently validated by Thermo Fisher Scientific and is not included within the kit's standard performance specifications. As such, users should evaluate suitability within their specific system and experimental conditions, as results may vary depending on the culture model and workflow.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormatKit
High-throughput CompatibilityYes
Isolation TechnologyDirect Cell Lysis
No. of Reactions40 Reactions
Product TypeDirect Cell Lysis to RT-qPCR Kit
Purification TargetRNA
Quantity40 Reactions
Sample TypeHepatocytes, HepaRG, HepG2
Shipping ConditionFrozen, Dry Ice
Starting MaterialUp to 100,000 Cultured Cells
TargetcDNA
Detection MethodTaqMan
For Use With (Application)Gene Expression
FormLiquid
GC-Rich PCR PerformanceHigh
Label or DyeROX
PCR Method2-step RT-qPCR
Reaction Speed10 min.
Unit SizeEach
Contents & Storage
HepatoExpress Lysis Solution (Room Temperature), HepatoExpress LysisGuard (-20°C), Express ezDNase (-20°C), SuperScript IV VILO Master Mix (-20°C), SuperScript IV VILO No RT Control (-20°C), TaqMan Fast Advanced Master Mix (4°C). For long-term storage, store HepatoExpress Lysis Buffer at 4°C.

Frequently asked questions (FAQs)

Is the TaqMan Cells-to-CT HepatoExpress Kit compatible with 3D cultures?

The TaqMan Cells-to-CT HepatoExpress Kit has not been tested with 3D cultures.

Is the TaqMan Cells-to-CT HepatoExpress Kit compatible with 384-well tissue culture plates?

The TaqMan Cells-to-CT HepatoExpress Kit has not been tested with tissue cells in 384-well plates.

Does the ezDNase included in the TaqMan Cells-to-CT HepatoExpress Kit need to be inactivated prior to RT-qPCR?

No, ezDNase contained in the TaqMan Cells-to-CT HepatoExpress Kit is a double-strand specific DNase and will not degrade cDNA produced in the RT reaction. It is also heat-labile and will be inactivated during the heat kill step of the RT reaction.

Should I wash my cells using warm or cold wash buffer when using the TaqMan Cells-to-CT HepatoExpress Kit?

With the TaqMan Cells-to-CT HepatoExpress Kit, cells can be washed using warm or cold wash buffer according to user preference. In our evaluation, we found that using warm or chilled wash buffer yielded equivalent results.

When using the TaqMan Cells-to-CT HepatoExpress Kit, can I go directly from cells to RT-qPCR without having to purify RNA?

Yes, the TaqMan Cells-to-CT HepatoExpress Kit allows for direct RT-qPCR amplification from cell lysates without purification of RNA.