Search
Search

| Catalog Number | Diameter (Metric) Inner | Length (Metric) | Bed Volume |
|---|---|---|---|
| A65886 | 5 cm | 5 cm | 100 mL |
| A65878 | 2.5 cm | 5 cm | 25 mL |
| A65879 | 2.5 cm | 10 cm | 50 mL |
| A65887 | 5 cm | 10 cm | 200 mL |
| A65888 | 8 cm | 5 cm | 250 mL |
| A65889 | 8 cm | 10 cm | 500 mL |
| A65890 | 10 cm | 5 cm | 390 mL |
| A65891 | 10 cm | 10 cm | 785 mL |
| A40011735 | 14 cm | 5 cm | 770 mL |
| A40011733 | 14 cm | 10 cm | 1540 mL |
| A40011730 | 20 cm | 5 cm | 1570 mL |
| A40011738 | 20 cm | 10 cm | 3140 mL |
POROS GoPure AAVX chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAVX columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards. Each column is accompanied by column qualification documentation including bioburden and endotoxin. Packed with POROS™ CaptureSelect™ AAVX affinity resin, POROS GoPure AAVX columns offer high specificity and capacity for a broad range of AAV viral vector serotypes over a wide range of flow rates.
Features of POROS CaptureSelect AAVX resin include one-step AAV purification from crude material with high purity, with high specificity and capacity, improving yield by reducing process steps. Free of animal-origin components, the CaptureSelect AAVX ligand is a 12 to 15 kDa single-domain antibody fragment produced in the yeast Saccharomyces cerevisiae.