POROS™ GoPure™ AAVX Column
Thermo Scientific™

POROS™ GoPure™ AAVX Column

POROS GoPure AAVX chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAVX columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDiámetro interior (métrico)Longitud (métrico)Volumen del lecho
A6589110 cm10 cm785 mL
A658782.5 cm5 cm25 mL
A658792.5 cm10 cm50 mL
A658865 cm5 cm100 mL
A658875 cm10 cm200 mL
A658888 cm5 cm250 mL
A658898 cm10 cm500 mL
A6589010 cm5 cm390 mL
A4001173514 cm5 cm770 mL
A4001173314 cm10 cm1540 mL
A4001173020 cm5 cm1570 mL
A4001173820 cm10 cm3140 mL
Número de catálogo A65891
Precio (MXN)
-
Diámetro interior (métrico):
10 cm
Longitud (métrico):
10 cm
Volumen del lecho:
785 mL

POROS GoPure AAVX chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAVX columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards. Each column is accompanied by column qualification documentation including bioburden and endotoxin. Packed with POROS™ CaptureSelect™ AAVX affinity resin, POROS GoPure AAVX columns offer high specificity and capacity for a broad range of AAV viral vector serotypes over a wide range of flow rates.

  • Pressure rating (bar/psi): 4/58
  • Operating temperature: 2°C to 30°C
  • Bed support size: 12 μm
  • Inlet/outlet internal diameter: 2.00 mm (2.5 cm), 3.45 mm (5 cm), 4.57 mm (8 cm), 6.35 mm (10 cm, 14 cm, 20 cm)
  • Inlet/outlet port connections: 3/4 inch mini tri-clamps
  • Includes a 30 mL resin side sample for incoming QC

Features of POROS CaptureSelect AAVX resin include one-step AAV purification from crude material with high purity, with high specificity and capacity, improving yield by reducing process steps. Free of animal-origin components, the CaptureSelect AAVX ligand is a 12 to 15 kDa single-domain antibody fragment produced in the yeast Saccharomyces cerevisiae.

For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to human or animals.
Especificaciones
Volumen del lecho785 mL
Longitud (imperial)4 in.
Cantidad1 Ea.
Diámetro interior (métrico)10 cm
Longitud (métrico)10 cm
Unit SizeEach