What is the difference between the CBQCA Plus Protein Quantitation Kit (Cat. No. A66522) and the CBQCA Protein Quantitation Kit (Cat. No. C6667)?
There are two main differences between the two kits. Firstly, the CBQCA Protein Quantitation Kit (Cat. No. C6667) contains a potassium cyanide (KCN) component (component C). This component has been removed in the CBQCA Plus Protein Quantitation Kit (Cat. No. A66522) and replaced with mandelonitrile which serves as a less toxic alternative to KCN without compromising assay performance. Secondly, the CBQCA Plus Protein Quantitation Kit includes one additional component compared to the CBQCA Protein Quantitation Kit, a BupH Borate Buffer Pack.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Quantitation Support Center.