Pierce™ Aquaporin Z Membrane Protein Standard
Thermo Scientific™

Pierce™ Aquaporin Z Membrane Protein Standard

The Thermo Scientific Pierce Aquaporin Z Membrane Protein Standard is a highly validated pure membrane protein formulated for direct infusion native mass spectrometry experiments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A666955 x 20 μg
A6669320 μg
Catalog number A66695
Price (EUR)
1.726,00
Each
Quantity:
5 x 20 μg
Price (EUR)
1.726,00
Each

The Thermo Scientific Pierce Aquaporin Z Membrane Protein Standard is a highly validated pure membrane protein formulated for direct infusion native mass spectrometry experiments.

Features of Pierce Aquaporin Z Membrane Protein Standard include:

  • Positive control sample—aquaporin Z protein production optimized and validated for use in native mass spectrometry
  • Rigorously tested—high quality, consistent protein standard documented via lot-specific certificate of analysis
  • Convenient—ready-to-use format with each single-use vial sufficient for 5 injections

Achieve confidence in your methods without wasting precious samples

Method development for membrane protein analysis provides unique challenges. Pierce Aquaporin Z Membrane Protein Standard empowers success by letting you test parameters and troubleshoot issues without wasting irreplaceable samples. This high purity standard contains sufficient material for multiple qualitative experiments by direct infusion or LC/MS. You can use it to verify system performance, tune parameters, and develop native LC/MS methods without worrying about depleting your real samples.

For Research Use Only.
Specifications
ColorColorless
DescriptionTetramer
For Use With (Equipment)Q-Exactive™ UHMR, HMRn option on Tribrid Series MS systems
FormulationLiquid in 0.05% LDAO, 200 mM Ammonium Acetate; pH 7.4
GradeMass Spec Grade
MaterialAquaporin Z
Molecular Weight (g/mol)98,120 Da
Quantity5 x 20 μg
FormLiquid
Product TypeProtein Standard
Unit SizeEach