Owl™ A6BP Wide Gel Horizontal Electrophoresis System
Owl™ A6BP Wide Gel Horizontal Electrophoresis System
Owl™ A6BP Wide Gel Horizontal Electrophoresis System
Thermo Scientific™

Owl™ A6BP Wide Gel Horizontal Electrophoresis System

The Thermo Scientific™ Owl™ A6BP Wide Gel Horizontal Electrophoresis System with buffer ports has been discontinued.
Have Questions?
Catalog NumberGel Size
A6BP23 x 25 cm
Catalog number A6BP
Price (USD)
-
Gel Size:
23 x 25 cm

The Thermo Scientific™ Owl™ A6BP Wide Gel Horizontal Electrophoresis System with buffer ports has been discontiued. Please consider the Owl A6 Wide Gel Horizontal Electrophoresis System as its replacement, an ideal system for high-throughput laboratories.

Features:
  • Screens 25 to 500 samples on a single agarose gel, producing clear, tight banding patterns with no “smiling.”
  • Unique comb-slot design of the gel tray allows use of up to 10 evenly spaced combs.
  • Using special combs and a multichannel pipetter, samples may be loaded directly from a 96, eight or 12-well plate.
  • Incorporates Owl's end-gate casting design—gel tray is fitted with gasketed end gates which provide a leakproof seal without tape.
  • Sample loading with multichannel pipetter speeds up transfer.
  • Bubble level ensures casting of flat, uniformly thick gels.

Includes:

  • Buffer chamber with three-point leveling base
  • Thermo Scientific™ SuperSafe™ lid with attached power supply leads
  • UVT gel tray with gasketed end gates
  • Four combs (two 25-well and two 50-well, 1.5mm thick)
Specifications
Certifications/ComplianceCE marked
DescriptionGel size: 23 x 25cm; Buffer volume: 3L; L x W x H: 35 x 38 x 11cm; With buffer ports
Dimensions (L x W x H)13.75 x 14.94 x 4.38 in. (35 x 38 x 11 cm)
Height (English)4.38 in.
Height (Metric)11 cm
Length (English)13.75 in.
Length (Metric)35 cm
Length (Metric) Gel25 cm
ModelModel A6 w/Buffer Ports
Sample Size25 to 500
Volume (Metric) Buffer3 L
Width (English)14.94 in.
Width (Metric)38 cm
Width (Metric) Gel23 cm
Gel Size23 x 25 cm
Product LineOwl
TypeWide
Unit Size-

Frequently asked questions (FAQs)

Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.