Actinomycin D, 10 mg - FAQs
Actinomycin D, 10 mg - FAQs
View additional product information for Actinomycin D - FAQs (A7592, 11805017)
1 product FAQs found
How should I reconstitute Actinomycin D?
We recommend reconstituting Actinomycin D in either DMF or DMSO; it should be soluble to at least 1 mM in either solvent.