Thermo Scientific™
Cytospin™ 4 Cytocentrifuge Accessories
Related applications:
Anatomical Pathology
Use these accessories with the Thermo Scientific™ Cytospin™ 4 Cytocentrifuge.
Back to top
  • Shorten preparation time by having a second sealed head ready when the first finishes processing
  • Autoclavable
  • 12-chamber head seals completely