Thermo Scientific™
Manual Staining Station, Manual staining station
Related applications:
Anatomical Pathology
Ideal for unique staining needs, frozen sections or special stains, Thermo Scientific™ Manual Staining Station uses the same staining troughs and slide baskets as the Varistain™ Gemini stainer.
Back to top
  • Sturdy cover closes over troughs to minimize evaporation.
  • Baskets are designed to lock slides into position even when turned upside down.
  • Slide baskets hold 20 standard microscope slides.
  • Troughs hold 11 fl. oz. (320mL) of reagent.
  • Best when used for low-volume procedures.
Physical Specifications
Dimensions (D x W x H): 7.1 x 39.4 x 4.7″ (18 x 100 x 12cm)
Includes:
Kit includes a steel support rack, a complete set of 14 staining troughs (320mL, No. A78010487) and five slide baskets with carriers (No. A78010466).