Thermo Scientific™
Blade Holder Packages for the Cryotome™ FSE Cryostat
Related applications:
Anatomical Pathology
Hold blades for the Cryotome™ FSE Cryostat with these Thermo Scientific™ blade holder packages.
Back to top
Universal Blade Holder:
  • Accepts standard C- or D-profile knife up to 220mm; 120mm is standard length
  • Use with standard steel knives or with disposable blade holder
  • Use in conjunction with disposable blade holders (Part Nos. 40 or 47)
  • Low-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder for use with low-profile disposable blades
  • High-Profile Blade Holder:
  • Lateral movement of blade holder eliminates need to unclamp and reclamp blade
  • Color coding identifies lever function
  • Holder will accept either standard or heavy-duty high-profile blades, providing a wide range of tissue sectioning options