Thermo Scientific™
Cryotome™ Cryostat Accessories
 Related applications:
Anatomical Pathology
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Thermo Scientific™ Cryotome™ Cryostat Accessories.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on active Peltier
  • Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples
Documents
Manuals & protocols