Thermo Scientific™
Cryotome™ Cryostat Accessories
Related applications:
Anatomical Pathology
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Thermo Scientific™ Cryotome™ Cryostat Accessories.
Back to top
  • Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
  • Freezing time is shortened if freezing stage and tissue are placed on active Peltier
  • Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples