Thermo Scientific™
Shandon™ ClearVue™ Coverslipper Coverslip Hoppers
 Related applications:
Anatomical Pathology
Designed for use with the Thermo Scientific™ Shandon™ ClearVue™ Coverslipper, the Thermo Scientific™ Coverslip Hoppers meets the needs of most laboratories.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Makes integration into the laboratory effortless
  • Ships preloaded with 500 or 575 slips each
  • Inserts quickly into the Shandon ClearVue coverslipper without workflow interruption
  • Requires no manual handling of glass
  • Dispenses a clean coverslip each and every time
Documents
Manuals & protocols