Acetonitrile, UHPLC-MS
Acetonitrile, UHPLC-MS
Acetonitrile, UHPLC-MS
Acetonitrile, UHPLC-MS
Acetonitrile, UHPLC-MS
Acetonitrile, UHPLC-MS
Thermo Scientific™

Acetonitrile, UHPLC-MS

Unique signal-to-noise specification for the chemical industry where the solvent quality is linked directly to the sensitivity of the mass spectrometer. | CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Catalog NumberQuantityPackaging
A9561CS
also known as A9561
1 LClear Borosilicate Glass Bottle
Catalog number A9561CS
also known as A9561
Price (EUR)
1.578,00
Case of 6
Quantity:
1 L
Packaging:
Clear Borosilicate Glass Bottle
Price (EUR)
1.578,00
Case of 6
Acetonitrile, C2H3N, CAS Number-75-05-8, cyanure de methyl, methanecarbonitrile, cyanomethane, acetonitrile, mecn, methane, cyano, methyl cyanide, ethanenitrile, methylkyanid, ethyl nitrile, 1L
RUO – Research Use Only
Specifications
Absorbance≤0.005 AU at 235nm, ≤0.005 AU at 254nm, ≤0.005 AU at 280nm, ≤0.01 AU at 230nm, ≤0.015 AU at 220nm, ≤0.015 AU at 225nm, ≤0.025 AU at 215nm, ≤0.03 AU at 210nm, ≤0.04 AU at 205nm, ≤0.05 AU at 200nm, ≤0.15 AU at 195nm, ≤1.00 AU at 190nm
Assay Percent Range99.9% minimum by GC
Boiling Point81°C
CAS75-05-8
ChEBICHEBI:38472
Chemical Name or MaterialAcetonitrile
ColorColorless
Density0.781 g/cm3
Flash Point−40°C (-40°F)
IUPAC Nameacetonitrile
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
Melting Point-46°C
Molecular FormulaC2H3N
PackagingClear Borosilicate Glass Bottle
Physical FormLiquid
PubChem CID6342
Recommended StorageRoom temperature
Residue after Evaporation0.8ppm max.
SMILESCC#N
Synonymmethyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Titratable Acid0.008meq/g max.
Titratable Base0.0006meq/g max.
Vapor Pressure97mbar at 20°C
Viscosity0.36 cP at 20°C
Water0.01% max.
GradeUHPLC-MS
Molecular Weight (g/mol)41.053
Quantity1 L
Unit SizeCase of 6