Notice: This product will be discontinued on September 1, 2017. For superior performance, we recommend DreamTaq PCR Master Mix (2X).

ThermoPrime 2x PCR Master Mix, with 1.5 mM MgCl2

Catalog number: AB0575DCB

Thermo Scientific™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific ThermoPrime Taq DNA Polymerase is a 94 kDa recombinant thermostable DNA polymerase obtained by high level expression of the Taq DNA polymerase gene in E. coli. A proprietary buffer component protects ThermoPrime DNA polymerase during high temperature steps, ensuring optimal performance throughout the reaction, leading to increased PCR yields. Convenient Master Mix format allows easier reaction set-up.

Highlights

• Enhanced enzyme stability for increased yield
• Advanced buffering system minimizes PCR optimization steps
• Convenient Master Mix formats save time and reduce the risk of sample contamination during assay setup

Applications

• Amplification of fragments up to 5 kb
• Routine PCR

Related Products
ThermoPrime 2x PCR Master Mix, with 1.5 mM MgCl2
ThermoPrime Taq DNA Polymerase, with 10x buffer and separate vial of 25 mM MgCl2
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Polymerase: ThermoPrime Taq DNA Polymerase
Hot Start: No
Fidelity (vs. Taq): 1 X
GC-Rich PCR Performance: Low
Reaction Format: SuperMix or Master Mix
Product Overhang: 3'-A
Reaction Speed: Standard
Product Line: ThermoPrime
Product Size: 1,600 reactions

Contents & storage

Contains:
• 20 x 1 mL PCR Master Mix with 1.5 mM MgCl2 (2X)

Store at -20°C.

Documents

Manuals & protocols

Recommended products