Abgene Storage Plate, 96-well, 0.8 mL, individually wrapped, black

Catalog number: AB0765K

Thermo Scientific™  Related applications: Cell Counting, Viability, & Cryopreservation | Microplates, Dishes and Flasks

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific ABgene 0.8mL Storage Plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored or transported easily. All plates are ANSI format for compatibility with automated systems. To further assist in storage and tracking, all plates can be supplied with custom barcoding.

Features of Thermo Scientific ABgene 0.8mL Storage Plate, Black

• 0.8 mL maximum well volume when sealed with adhesive or heat seals
• Conical well bottom for ease of sample retrieval• Individually wrapped for convenience
• Polypropylene
• Black plates are ideal for fluorescence-based assays.
• Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
• Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
• Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
• Cleanroom manufactured
• ANSI-format for compatibility with automated systems

Applications
• Sample storage
• Sample resuspension
• Cell culture
• Assay setup: diluting and aliquoting

Related Products
Abgene Storage Plate, 96-well, 0.8 mL, individually wrapped
Abgene Storage Plate, 96-well, 0.8 mL, individually wrapped, white
ABgene Storage Plate, 96-well, 0.8 mL, bulk packed
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: 96-well plate
Plate Type: Storage Plate
Product Size: 50 plates
Shipping Condition: ELMS

Documents

Manuals & protocols