Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates
Thermo Scientific™

Abgene™ 96 Well Polypropylene Sample Processing & Storage Microplates

Streamline your molecular biology applications with these 96-well Polypropylene Storage Plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell ShapeVolume (Metric) Well
AB0796Round330 μL
AB1058
also known as AB-1058
Round, Conical Bottom (V-well)200 μL
Catalog number AB0796
Price (USD)
257.86
Online exclusive
321.76
Save 63.90
Case of 100
Well Shape:
Round
Volume (Metric) Well:
330 μL
Price (USD)
257.86
Online exclusive
321.76
Save 63.90
Case of 100
Thermo Scientific Abgene 96 Well Polypropylene Storage Microplates provide excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. All plates are ANSI format for compatibility with automated systems and are manufactured in our cleanroom facility to ensure molecular grade quality. To further assist in storage and tracking, all plates can be supplied with custom barcoding.

Features

  • Alphanumeric grid-referencing and industry standard footprint
  • Polypropylene
  • Sealing options: Adhesive Seals, Heat Seals and Cap Strips
  • Cleanroom manufactured

Applications

  • Sample storage
  • Sample resuspension
  • Cell culture
  • Assay setup: diluting and aliquoting
Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Dnase FreeYes
Description96-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
FormatStandard
Max. RCF5000 x g
No. per Case100
No. per Pack1
PackagingBulk
Plate SizeANSI/ SLAS Standard
Product Type96-well plate
RNase FreeYes
Regulatory StatusRUO
Volume (Metric) Well330 μL
Volume (Metric) Working300 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorNatural
Fit for PurposePCR Ready
MaterialPolypropylene
No. of Wells96
Product LineAbgene
SterilitySterile
Surface TreatmentNon-treated
Well ShapeRound
Unit SizeCase of 100

Frequently asked questions (FAQs)

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What 96-well plates do you recommend for -80 degrees C storage?

Both polypropylene and polystyrene plates can be used to store samples at -80 degrees C and depends on the downstream application. Please note that the sealing method selected is critical for storing at these temperatures and heat sealing is recommended.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.