PCR Plate, 96-well, low profile, skirted, 150 plates - FAQs
PCR Plate, 96-well, low profile, skirted, 150 plates - FAQs
View additional product information for PCR Plate, 96-well, low profile, skirted - FAQs (AB0800150, AB0800)
2 product FAQs found
What is the difference between PCR Plate, 96-well, low profile, skirted (Cat. No. AB0800, AB0800150) and PCR Plate, 96-well, robotic (Cat. No. AB1300)?
There is a slight difference in the height of the PCR plates. PCR Plate, 96-well, low profile, skirted (Cat. No. AB0800, AB0800150) is 15.5 mm tall and PCR Plate, 96-well, robotic (Cat. No. AB1300) is 17.0 mm tall.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
What is the PCR Plate, 96-well, low profile, skirted (Cat. No. AB0800, AB0800150) made of?
The PCR Plate, 96-well, low profile, skirted is made of polypropylene.