Separation Plate, 96-well

Catalog number: AB0952

Thermo Scientific™  Related applications: Cell Counting, Viability, & Cryopreservation | Microplates, Dishes and Flasks

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Ready-to-use microplate format

• 0.6mL well volume
• Contains polyethylene frits
• Pore size 30-45µm
• Suitable for centrifugation
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Format: 96-well plate
Plate Type: Filter Plate
Product Size: 24 plates
Shipping Condition: ELMS

Documents

Manuals & protocols