Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
PCR Plate, 96-well, rigid, semi-skirted
Thermo Scientific™

PCR Plate, 96-well, rigid, semi-skirted

The Thermo Scientific 96-Well Thermo–Fast Semi-Skirted Plates, Segmented provides superior rigidity to the plates and provides space for barcoding. TheRead more
Have Questions?
Catalog NumberQuantity
AB099025 Plates
Catalog number AB0990
Price (USD)
199.00
Each
Quantity:
25 Plates
Price (USD)
199.00
Each
The Thermo Scientific 96-Well Thermo–Fast Semi-Skirted Plates, Segmented provides superior rigidity to the plates and provides space for barcoding. The patented segmented design of this plate allows it to be cut into 24 and 48 well sections.

Features of The Thermo Scientific Thermo–Fast 96-Well Semi-Skirted Plates, Segmented:

• Patented segmented plate design—allows plates to be cut into 24 and 48 well sections
• Semi skirt—adds rigidity and allows for labeling or barcoding
• Raised rim—around each well enables secure sealing and safeguards against evaporation
• 0.3 mL maximum well volume—when used with adhesive and heat seals
• Uniform, thin well walls—deliver maximum and consistent heat transfer
• White plates—optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: H1

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
Fast PCR Plate, 96-well, low profile, semi-skirted, raised deck, barcoded
Fast PCR Plate, 96-well, low profile, semi-skirted, raised deck, white, barcoded
Specifications
Certifications/ComplianceCertified RNase/DNase-free
For Use With (Equipment)Real-Time PCR Systems or Thermal Cyclers
Packaging25 Plate
Product TypePCR Microplate
Quantity25 Plates
Reaction SpeedStandard
Skirt StyleSemi
Volume (Metric) Well0.3 mL
Volume (Metric) Working0.2 mL
BarcodeNo Barcode
ColorClear
MaterialPolypropylene
No. of Wells96
Unit SizeEach