Notice: This product will be discontinued on May 1, 2017. For superior performance, we recommend the MicroAmp® Optical 96-Well Reaction Plate.

PCR Plate, 96-well, semi-skirted, raised deck, yellow

Catalog number: AB1100Y

Thermo Scientific™  Related applications: PCR

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The Thermo Scientific 96-Well Semi-Skirted Plates, Raised Deck, Yellow, are specifically designed to be directly compatible with all ABI platforms, including 96-well sequencers, without the need for adapters. The plates have a cut corner at the A12 location and are available with barcodes or as a SuperPlate for robotic handling. The bottom of the well is clear for better sample visibility.

Features of The Thermo Scientific 96-Well Semi-Skirted Plates, Raised Deck

• 0.3 mL maximum well volume (when used with adhesive and heat seals)
• Yellow color to aid in sample tracking
• A12 cut corner
• Clear well bottom for sample visibility
• Available with barcode
• Sealable using Thermo-Mats, Adhesive Films and Foils, Heat Seals, and Flat, Domed, and Ultra Clear Cap Strips
• Available as a SuperPlate providing 4X more rigidity for superior robotic handling
• Also available in opaque black and white for fluorescent and luminescent based procedures. Other colors are also available

Applications

• PCR

Related Products
PCR Plate, 96-well, semi-skirted, raised deck
PCR Plate, 96-well, semi-skirted, raised deck, white
PCR Plate, 96-well, semi-skirted, raised deck, blue
PCR Plate, 96-well, semi-skirted, raised deck, green
PCR Plate, 96-well, semi-skirted, raised deck, black
PCR Plate, 96-well, semi-skirted, raised deck, purple
PCR Plate, 96-well, semi-skirted, raised deck, red
SuperPlate PCR Plate, 96-well, semi-skirted, raised deck
SuperPlate PCR Plate, 96-well, semi-skirted, raised deck, white
PCR Plate, 96-well, semi-skirted, raised deck, barcoded, white
PCR Plate, 96-well, segmented, semi-skirted
PCR Plate, 96-well, segmented, semi-skirted, white
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Barcode: No Barcode
Color: Yellow
For Use With (Equipment): Thermal Cyclers
Format: 96-well plate, Semi-skirted
Product Size: 25 plates
Reaction Speed: Standard

Documents

Manuals & protocols