Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate
Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate

Abgene™ 96 Well 1.2 mL Polypropylene DeepWell™ Sample Processing & Storage Plate

Optimize suspension cell culture with these U-bottomed 1.2 mL storage plates.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberWell ShapePackaging
AB1127Square, Round Bottom (U-well)Individually Wrapped
AB0564RoundIndividually Wrapped
AB0787RoundBulk
Catalog number AB1127
Price (USD)
287.75
Online exclusive
431.38
Save 143.63
Case of 50
Well Shape:
Square, Round Bottom (U-well)
Packaging:
Individually Wrapped
Price (USD)
287.75
Online exclusive
431.38
Save 143.63
Case of 50

Thermo Scientific™ Abgene™ polypropylene microplates and deepwell plates offer storage security for assays, compound libraries or storing samples for either intermediate or long term use. Abgene microtiter plates are manufactured to exacting specifications in our Class 100,000 clean room ISO 9001 conditions using high quality medical grade virgin polypropylene resins, which ensure confidence in the quality and performance of the storage plates. Designed to ANSI standards these microplates and deepwell plates are compatible with a variety of automated liquid handling for high throughput workflows. The polypropylene storage plates are offered with spindle, pyramidal, U and V bottom wells along with a wide range of volumes to accommodate most applications.

Features

  • Round well shape designed for optimal sample recovery or square shape to maximize sample volume within ANSI footprint design
  • Each well has an independent sealing rim to prevent cross-contamination
  • U-bottomed wells ideally suited for sample resuspension
  • Polypropylene (AB-1068, polystyrene)
  • Resistant to DMSO (100%); Ethanol (100%); Isopropanol (100%)
  • Suitable for Autoclaving (15 minutes at 121°C) or Gamma Irradiation
  • Sealing options: Adhesive Seals, Heat Seals, Storage Plate Caps and Cap Strips, and Storage Plate Sealing Mats
  • Cleanroom manufactured
  • ANSI-format for compatibility with automated systems
Specifications
AutoclavableAutoclavable
Binding TypeLow binding PP
Certifications/ComplianceCertified RNase, DNase, PCR Inhibitors, and human DNA Free. ANSI Compliant
Dnase FreeYes
Description96-well Polypropylene Sample Processing & Storage Plates
For Use With (Application)Automation Liquid Handlers, Biobanking, Genomics, Sample Prep, NGS, Sequencing, Long and Short Term Storage, Drug Discovery, Vaccine Research, PCR and qPCR sample preparation
For Use With (Equipment)ALPS5000, ALPS3000, and ALPS50V Heat Sealers, Automated Liquid Handlers, Multidrop Combi, Centrifuges, Ultralow Freezers, Liquid Handlers
FormatStandard
Max. RCF2000 x g
No. per Case50
No. per Pack1
PackagingIndividually Wrapped
Plate SizeANSI/ SLAS Standard
Product Type96-well plate
RNase FreeYes
Regulatory StatusRUO
Volume (Metric) Well1200 μL
Volume (Metric) Working1000 μL
Array8 x 12
BarcodeCustom Barcoding Available
ColorNatural
Fit for PurposePCR Ready
MaterialPolypropylene
No. of Wells96
Product LineAbgene
SterilitySterile
Surface TreatmentUntreated
Well ShapeSquare, Round Bottom (U-well)
Unit SizeCase of 50

Frequently asked questions (FAQs)

Are Thermo Scientific ABgene plastics endotoxin-free?

They are not certified endotoxin-free. However, their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Are Thermo Scientific ABgene plastics sterile?

Thermo Scientific Abgene plastics are not typically sterilized as their entire production process, from molding to final packaging, is carried out in a Class 100,000 clean room under ISO 9001 guidelines. All Thermo Scientific PCR plastics are certified free from RNase, DNase and human DNA. In contrast, during typical non-clean room production, plastics are exposed to many contaminants including dust, bacterial cells, and DNA. The plastics are then sterilized to kill bacteria and inactivate RNases and DNases, but sterilization does not remove dust or DNA contamination. The dust particles left behind can inhibit PCR, and the damaged DNA fragments can still act as templates, leading to non-specific amplification.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

What 96-well plates do you recommend for -80 degrees C storage?

Both polypropylene and polystyrene plates can be used to store samples at -80 degrees C and depends on the downstream application. Please note that the sealing method selected is critical for storing at these temperatures and heat sealing is recommended.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.