The Thermo Scientific Verso 1-step RT-qPCR ROX Mix Kit for probe chemistry offers a quick and simple way to detectRead more
Have Questions?
Change view
Catalog Number
Quantity
AB4101A
also known as AB-4101/A
200 x 25 μL Reactions
AB4101C
also known as AB-4101/C
400 x 25 μL Reactions
2 Options
Catalog number AB4101A
also known as AB-4101/A
Price (USD)
688.00
Each
Quantity:
200 x 25 μL Reactions
Price (USD)
688.00
Each
The Thermo Scientific Verso 1-step RT-qPCR ROX Mix Kit for probe chemistry offers a quick and simple way to detect mRNA. The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures.
Features of the Verso 1-step RT-qPCR ROX Mix Kit:
• Eliminate the need for DNase treatments—RT Enhancer (optional) prevents gDNA carry-over • RT and qPCR steps performed in single tube—convenience and reduced contamination risk • Use of the entire RT reaction as template for qPCR—improved sensitivity • Verso RT enzyme is highly sensitive with broad dynamic range—accurate detection of RNA from 1 pg-1 ug • Verso RT enzyme has a wide working temperature range (42 to 57°C)—improved success with GC-rich and other difficult templates • Contains an inert blue dye for ease of pipetting during reaction set up
An optional RT Enhancer is also provided and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment. All Verso 1-step RT-qPCR kits incorporate an inert dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.
In a 1-step approach to RT-qPCR, both the RT step and qPCR steps are performed in a single tube (1-step RT-qPCR). Adopting a 1-step approach is ideal when no cDNA storage is necessary for subsequent experiments. The approach minimizes the risk of contamination and improves reproducibility by reducing sample handling.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
FormatTube
No. of Reactions200 Reactions
Product LineVerso
Product Type1-Step QRT-PCR Kit w/ROX
Quantity200 x 25 μL Reactions
Sample TypeRNA
Sufficient For200 Reactions
Detection MethodPrimer-probe
For Use With (Application)Gene Expression, Pathogen Detection
GC-Rich PCR PerformanceHigh
PCR Method1-step RT-qPCR
Unit SizeEach
Contents & Storage
Contains: • 1 x 50 μL Verso Enzyme Mix • 1 x 250 μL RT enhancer • 2 x 1.25 mL 2X 1-Step qPCR ROX Mix Sufficient for 200 x 25 μL reactions.
Store at -20°C
Frequently asked questions (FAQs)
What is the RT Enhancer that is included in all Verso kits?
The RT Enhancer is a unique enzyme that degrades contaminating genomic DNA at the start of the RT step. It is equally as effective as DNase I treatment but more convenient as it eliminates the need for a separate DNase I incubation prior to RT-PCR.
Do I need to treat my RNA with DNase I before running RT-qPCR with Verso cDNA Synthesis Kit?
No, you do not need to treat your RNA with DNase I before running RT-qPCR with the Verso cDNA Synthesis Kit. The RT enhancer included in the Verso kit degrades any contaminating double-stranded DNA during the reverse transcription process, eliminating the need for a separate DNase I treatment.